TLC Analysis of Sugars on Silica gel 60 NH2
TLC Analysis of Sugars on Silica gel 60 NH2
Conditions
plate 1.13192 HPTLC Silica gel 60 NH2 20 cm x10cm (1.13192.0001)
eluent water/acetonitrile (30/70) for 55 mm followed by drying the plate;
(then ethylacetate/pyridine/water/glacial acetic acid/propanoic acid
(50/50/10/5/5) for 75 mm followed by drying the plate )
migration distance 55 mm (eluent 1), 75 mm (eluent 2)
chamber normal chamber without chamber saturation
sample 2 μL
detector Heat 5 min at 150 °C Detection
Description
suitability application for TLC