TLC Analysis of Water Soluble Vitamins on HPTLC Silica Gel 60 F 254 with Concentrating Zone
TLC Analysis of Water Soluble Vitamins on HPTLC Silica Gel 60 F 254 with Concentrating Zone
Conditions
plate  TLC pre-coated plate silica gel 60 F 254 with concentrating zone
(1.13728.0001, 1.13727.0001)
eluent [A] chloroform; [B] methanol; (60:40, A:B) 80% saturated with water,
containing 01. M lithium chloride
migration distance  6 cm
chamber  normal chamber without chamber saturation
sample  1 μL
detector  in situ evaluation with TLC/HPTLC scanner (Camag) 270 nm
Description
suitability application for TLC
Featured Industry Vitamins, Nutraceuticals, and Natural Products