TLC Analysys of Compounds in Saraca dives Fabaceae extract on HPTLC Silica gel 60 F254s
TLC Analysys of Compounds in Saraca dives Fabaceae extract on HPTLC Silica gel 60 F254s
Conditions
sample preparation 50 mg dried Extract in 10 mL Methanol was filtered over a 0.45μm syringe filter.
Application by CAMAG ATS4 6 mm bandwise.
plate 1.15696 HPTLC Silica gel 60 F254s 20 cm x10 cm (1.15696.0001)
eluent [A] ethylacetat; [B] formic acid; [C] glacial acid; [ D] water; (100:11:11:27, A:B:C) containing 0,1% n-Heptane
migration distance 5 cm
chamber normal chamber without chamber saturation
detector 366 nm Diphenylboric acid-2-aminoethylester-reagent
suitability application for TLC
Featured Industry Chemicals and Industrial Polymers
Description
suitability application for TLC