Attention:
We are currently upgrading the certain systems that support this website.  You may experience some functionality loss, though we expect the site to be fully functional by 9:00PM CT (US), February 24th.
We apologize for any inconvenience and appreciate your patience during this improvement process.Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Adsorbent Selection Guide
In addition to the general guidelines presented in the tables below view the technical report titled “A Tool for Selecting an Adsorbent for Thermal Desorption Applications” (1,770Kb pdf - T402025), which highlights the relative differences between the most commonly used adsorbents with the help of color-coded performance charts.
General Guidelines for Choosing Carbon Adsorbents
For multi-bed tubes, use the weaker adsorbent in front of the stronger adsorbent. For example, use Carbopack™ C in front of Carbopack B.
Relative Analyte Size1 Recommended Materials (listed weakest to strongest)
> C20 Carbotrap™ F, Carbopack F
C12-C20 Carbotrap C, Carbopack C, Carbotrap Y, Carbopack Y
C5-C12 Carbotrap B, Carbopack B
C3-C9 Carboxen™ 1016, Carbotrap X, Carbopack X, Carbopack Z
C2-C5 Carboxen 569, Carbosieve™ G, Carboxen 1000, Carbosieve S-III, Carboxen 1021, Carboxen 1018, Carboxen 1003, Carboxen 1012
1Analyte size relative to n-Alkanes. Consider all atoms, not just Carbon. For example, even though 1,2-Dichloroethane is a C2, the two Chlorine atoms give it a relative size between C4 and C5.
back to top
Physical Characteristics of Supelco Carbon Adsorbents
Adsorbent BET
surface area1
(m2/g)
Density
(g/mL)
Porosity (cc/g) Micropore
Diameter
(Å)
micro- meso- macro-
Carbotrap graphitized carbon black (20/40 mesh )
Carbotrap F 5 0.69 - - - -
Carbotrap C 10 0.68 - - - -
Carbotrap Y 24 0.45 - - - -
Carbotrap B 100 0.37 - - - -
Carbotrap X 240 0.43 - 0.62 - 100
Carbopack graphitized carbon black (60/80 mesh)
Carbopack F 5 0.64 - - - -
Carbopack C 10 0.68 - - - -
Carbopack Y 24 0.42 - - - -
Carbopack B 100 0.35 - - - -
Carbopack Z 220 0.18 - 1.73 - 255
Carbopack X 240 0.41 - 0.62 - 100
Carbon Molecular Sieve
Carboxen-1016 75 0.40 - 0.34 - -
Carboxen-569 485 0.58 0.20 0.14 0.10 5 - 8
Carboxen- 10212 600 0.62 0.30 - - 5 - 8
Carboxen-10182 675 0.60 0.35 - - 6 - 8
Carbosieve S-III3 975 0.61 0.35 0.04 -  4 - 11
Carboxen-1003 1000 0.46 0.38 0.26 0.28 5 - 8
Carbosieve G 1160 - 0.49 0.02 -  6 - 15
Carboxen-1000 1200 0.48 0.44 0.16 0.25 10 - 12
Carboxen-1012 1500 0.50 - 0.66 - 19 - 21
1 Brunauer, Emmett, Teller (BET) surface area calculations
2 microporous, monoporous carbon sieve
3 closed pore structure
back to top
Adsorbent Cross Reference
Adsorbent Supelco Equivalent or Substitute
Ambersorb® 347    Carboxen 564
Anasorb C300 Hopcalite
Anasorb CMS Carboxen 564, Carboxen 1000, Carbosieve S-III
Anasorb CSC Coconut charcoal
Anasorb GCB1 Carbopack B, Carbotrap B
Anasorb GCB2 Carbopack C, Carbotrap C
Anasorb 708 Chromosorb® 108
Anasorb 727 XAD-4
Anasorb 747 Carboxen 564, Carboxen 1000, Carbosieve S-III
Carbograph 1 Carbopack B, Carbotrap B
Carbograph 2 Carbopack C, Carbotrap C
Carbosphere Carboxen 1000
Graphpac-GC Carbopack B, Carbotrap B, Carbopack C Carbotrap C    
Hydrar Hopcalite
Purosieve Carboxen 1000
Spherocarb Carboxen 1000
Tenax® GC Tenax® TA