Absorption & Filtration Materials
Dependable, flexible and ecological solutions
Absorption, adsorption and filtration are among the most important applications in laboratory practice. We offer an extensive portfolio of reagents and materials with excellent absorption properties for a wide range of substances. Our high-quality products are available in a variety of pack sizes to suit your individual needs.
Absorption & filtration portfolio
  • Absorption tubes
  • Calcium oxide
  • Activated charcoal
  • Graphite
  • Molecular sieves
  • Soda lime
  • Aluminum oxide fibers
  • Glass wool
  • Sea sand
Protecting our customers & environment
Many of our absorption and filtration materials are produced from environmentally friendly, natural raw materials, such as our charcoal products or calcium oxide. Another example is our aluminum oxide fibers, which can be used as filtration and insulation agents due to their high melting point of 1,700 to 1,800 °C. Unlike asbestos fibers, they are not carcinogenic, and can be easily and safely disposed of.
Benefits
  • High-performance: Excellent take-up capacity 
  • Reliable: High quality for accurate results 
  • Flexible: Large choice of pack sizes
  • Ecological: Many natural raw materials
Materials
If you have trouble finding the products you're looking for, please contact our customer support for assistance.