Hydrofluoric Acid Passport
Carry with You
Hydrofluoric acid is one of the most dangerous acids known. Working with hazardous chemicals such as hydrogen fluoride, hydrofluoric acid and inorganic fluorides requires special provisions for the first aid treatment. Every accident involving these substances can have dramatic consequences for the injured person.
Chemical burns from hydrogen fluoride, hydrofluoric acid and inorganic fluorides can be life threatening if the poisonous effects of the fluoride ions are not treated.
Not every physician has the necessary specialist knowledge to properly treat accidental HF exposure. Therefore, we provide a credit card-sized HF Passport, which sets out the correct medical treatment. Everyone who works with these materials should carry this HF passport.
Download the Hydrofluoric Acid Passport (PDF, 0.2 MB)
Secure Dosing Aid
To avoid chemical burns from hydrofluoric acid, we offer our hydrofluoric acid products in 500mL bottles with an innovative and secure dosing aid.
The special design of the dosing aid allows for drip-free pouring with drop-by-drop precision. In addition, we also offer our S40-Closure System for hydrofluoric acid, so that the bottles close securely and spills are prevented.
Benefit from our secure solutions for your daily lab work.
Ordering information for hydrofluoric acid in 500 mL HDPE bottles with dosing aid: