Immobilon®-P PVDF membrane for Western blotting
Immobilon®-P is the original, and most commonly used, PVDF transfer membrane for Western blotting. Available in sheets, rolls, and pre-cut Western blotting sandwiches.
  • Broad use 0.45 µm pore size transfer membrane
  • Recommended for most Western blotting, especially proteins >20 kDa
  • Stronger protein signals due to high adsorption and retention of protein in the membrane
  • Prolonged shelf life due to higher tensile strength
  • Easier stripping & reprobing with PVDF membranes
Materials