Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Steriflip® Filters
Fast, easy sterilization of cell culture media and reagents
Steriflip filters eliminate liquid transfer and speeds sample handling by integrating a filter into the mouth of a sterile tube to which the original sample tube is directly attached.
The Steriflip® filter’s revolutionizing design has made it a popular tool among cell culturists aiming to eliminate contamination while simultaneously increasing experimental throughput. Filtration using the Steriflip® device is straightforward, requiring only three steps:
  1. Attach filter to 50 mL sample tube
  2. Flip assembly upside down
  3. Apply vacuum and recover sample in new tube
Units with fast flow and low protein binding Millipore Express (PES) membrane
Product No. Description Pore Size Pack Size Add to Cart
SCGP00525
Steriflip-GP, 0.22 µm, polyethersulfone, gamma irradiated 0.22 µm 25
Each box of 25 filter units includes one centrifuge tube stand
Units with very low protein binding Durapore (PVDF) membraneClear Sorting & Filtering
Product No. Description Pore Size Pack Size Add to Cart
SE1M003M00 Steriflip-HV, 0.45 µm, PVDF, radio-sterilized 0.45 µm 25
SE1M179M6 Steriflip-GV, 0.22 µm, PVDF, radio-sterilized 0.22 µm 25
Each box of 25 filter units includes one centrifuge tube stand
Unit with nylon net filterClear Sorting & Filtering
Product No. Description Pore Size Pack Size Add to Cart
SCNY00020 Steriflip 20 µm, nylon net, radio-sterilized 20.0 µm 25
SCNY00040 Steriflip 40 µm, nylon net, radio-sterilized 40.0 µm 25
SCNY00060 Steriflip 60 µm, nylon net, radio-sterilized 60.0 µm 25
SCNY00100 Steriflip 100 µm, nylon net, radio-sterilized 100.0 µm 25
Each box of 25 filter units includes one centrifuge tube stand
Accessories
Product No. Description Pack Size Add to Cart
SC50FL025 Steriflip Funnel Attachment, non-sterile 25