Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
VEGF Pathway
Background
Formation of new blood vessels, a process known as angiogenesis, is a fundamental biological process required for embryonic development, tissue repair, ovulation and menstruation. Angiogenesis involves sprouting of new blood vessels from the existing blood vessels through the proliferation, migration and invasion of endothelial cells. This process is tightly regulated by pro- and anti-angiogenic factors, dysregulation of which can lead to abnormal vasculature. Several signaling mechanisms function to form new blood vessels, including vascular endothelial growth factors (VEGFs)/VEGFRs, angiopoietin/Tie receptors1, Platelet-derived growth factors (PDGFs)/PDGFRs2 and EphirinB2/EphirinB43 . Among these, VEGF signaling has been often recognized as the key regulator of angiogenesis.
VEGF and VEGF Receptors
Vascular permeability factor (VPF) was initially known to show hyperpermeability of tumor blood vessels and involved in formation of tumor-associated ascites4. Later, studies revealed vascular endothelial growth factors (VEGF) as an angiogenic growth factor displaying high specificity for endothelial cells5. Subsequently, it was discovered that both VEGF and VPF are encoded by a single VEGF gene and are products of alternate splicing. Analysis of VEGF/VPF cDNA clones also revealed significant homology to platelet-derived growth factor (PDGF).
Ligands
There are five structurally related VEGF ligands: VEGFA, VEGFB, VEGFC, VEGFD and Placenta Growth Factor (PIGF). All the VEGFs are homodimers with disulphide bonds. The native VEGF is heparin-binding homodimeric glycoprotein closely corresponds to the VEGF165 variant. VEGFA was shown to have five isoforms with 121, 145, 165, 189 and 206 amino acids as a result of alternate splicing6. VEGF ligands are secreted by various cell types and act in an autocrine and paracrine manner. Each VEGF ligand binds differently to the receptors and induces distinct biological responses7.
Receptors
VEGF ligands bind to three receptor tyrosine kinases, VEGFR1 (Flt-1), VEGFR2 (Flk-1, KDR) and VEGFR3 and to co-receptors like Neuropilin 1 (NRP1). While VEGF receptors have homology in kinase domains their signaling cascade differs significantly. Ligand binding induces receptor dimerization (homo/hetero dimers) and activation of the kinase domain.
Phylogenetic analyses revealed that D-VEGFR/PVR, found in Drosophila melanogaster is the common ancestor of the vertebrate VEGF receptors. Both VEGFR and PDGFR in vertebrates were most likely obtained from the duplication/triplication of single D-VEGFR/PVR gene8,9.
Product # V7259 V4512 SRP4363 SRP4364 SRP4365 V5765 SRP3182 H9166
Product VEGF VEGF VEGF165 VEGF165 VEGF165 VEGF165 VEGF VEGF165
Species Human Mouse Human Mouse Rat Human Human Human
Expressed in E. coli E. coli E. coli E. coli E. coli HEK E. coli HEK
Endotoxin-tested Yes Yes Yes Yes Yes Yes Yes Yes
Carrier-free No No No No No No Yes Yes
Purity ≥98% ≥98% ≥98% ≥98% ≥98% ≥95% ≥98% ≥95%
ED50/EC50 1-10 ng/mL 1-10 ng/mL ≤0.1 ng per ug ≤0.1 ng per mg ≤0.1 ng per ug ≤10 ng/mL 1.0-8.0 ng/mL 2-10 ng/mL
VEGF – Signal Transduction
VEGF signaling involves the cascade of signals triggered when the ligand binds the VEGF receptors. The downstream signaling includes activation of phospholipase Cγ1, MAPK pathway via Ras/Raf1 activation and PI3K/Akt pathway. Phospholipase Cγ1 regulates the concentration of intracellular Ca+2 ions and formation of endothelial nitric oxide synthase. The effect of all the cascades provides a balance of pro- and anti-angiogenic signals that maintain the vasculature and/or result in sprouting of new blood vessels, cell proliferation and cell migration (Figure 1). Table 1 summarizes signaling transduction through VEGF receptors.
Receptor VEGFR1 VEGFR2 VEGFR3 NRP1
Ligands VEGFA, VEGFB,PIGF VEGFA, VEGFC,VEGFD VEGFC,VEGFD VEGF165, PlGF
Cell Expression Endothelial, Dendritic, Monocytes/ Macrophages, Osteoblasts, Pericytes, Trophoblasts Endothelial, Neuronal, Retinal progenitors, hematopoietic stem cells,  Osteoblasts, Megakaryocytes Lymphatic endothelial cells, Monocytes/Macrophages Endothelial, Neuronal, Plasmacytoid Dendritic cells
Signal Transduction Activation of phospholipase C-γ1; regulation MAPK pathway10,11 Activation of PI3K/Akt pathway, BAD, FKHR1, Caspase1, Bcl-2 Activation of ERK and PI3K/Akt pathways Activation of MAPK pathway
Functions Inhibition of angiogenesis and recruitment of immune cells Angiogenesis; cell survival, proliferation, migration, vascular permeability and inhibition of apoptosis Lymphangiogenesis; development of vasculature; migration of endothelial cells Vascular development; apoptosis inhibition; migration
Table 1: Signaling transduction by VEGF receptors
VEGF signaling pathway (VEGF/VEGF receptor system)
Image 1: VEGF Pathway 
Factors Regulating VEGF and VEGFR Expression
There are several factors regulating the expression of VEGF and VEGFR. They are broadly classified into external factors, transcription factors and oncogenes. External factors such as hypoxia in tissue microenvironment regulate VEGF expression through Hypoxia Inducible Factor-1 alpha (HIF-1α)12. Under hypoxic conditions HIF-1α dimerizes with HIF-1β, binds to the VEGF promoter inducing VEGF transcription. Growth factors and cytokines secreted in tissue microenvironment such as epidermal growth factor receptors (ErbB1 and ErbB2), insulin like growth factor-I receptor (IGF-IR)13, hepatocyte growth factors14, platelet-derived growth factors (PDGF)15 and cyclooxygenases (COX)16 induce angiogenesis. Oncogenes and tumor suppressors such as like c-SRC (proto-oncogene)17, BCR-ABL18, Ras19, p5320 and PTEN21 also regulate the expression of VEGF.
VEGFR in Human Diseases
Diseases that are associated with vascular abnormalities are related to anomalies in VEGF/VEGFR signaling axis. Anti-VEGF treatment is being employed to treat certain type of cancers, and condition like  preeclampsia, macular degeneration and amyotrophic lateral sclerosis.22-28 The efficacy of antiangiogenic therapy for tumors is often met with challenges such as drug resistance and factors such as increased expression of HIF-1alpha which leads to increase in the number of cancer stem cells in the tumor niche.
Conclusion
The role of VEGF and VEGFRs is well recognized in angiogenesis and has been implicated in chronic disease conditions. Despite the development of small molecules and antibodies that suppress angiogenesis, several tumors showed resistance to anti-VEGF therapy due to parallel angiogenesis signals from other growth factors. Factors that contribute to tumor resistance and angiogenesis are yet to be extensively defined.
Explore complete offering of high quality VEGF recombinant proteins and receptors for your research use here.
Study VEGF signaling with consistency in your stem cell cultures. Visit our Stem Cell Learning Center to learn more.
 References
  1. Davis, S., Aldrich, T. H., Jones, P. F., Acheson, A., Compton, D. L., Jain, V., Ryan, T. E., Bruno, J., Radziejewski, C., Maisonpierre, P. C., and Yancopoulos, G. D. (1996) Isolation of angiopoietin-1, a ligand for the TIE2 receptor, by secretion-trap expression cloning. Cell 87, 1161–1169.
  2. Heldin, C. H., and Westermark, B. (1999) Mechanism of action and in vivo role of platelet-derived growth factor. Physiol. Rev. 79, 1283–1316.
  3. Wang, H. U., Chen, Z. F., and Anderson, D. J. (1998) Molecular distinction and angiogenic interaction between embryonic arteries and veins revealed by ephrin-B2 and its receptor Eph-B4. Cell 93, 741–753.
  4. Senger, D. R., Galli, S. J., Dvorak, A. M., Perruzzi, C. A., Harvey, V. S., and Dvorak, H. F. (1983) Tumor cells secrete a vascular permeability factor that promotes accumulation of ascites fluid. Science 219, 983–985.
  5. Leung, D. W., Cachianes, G., Kuang, W. J., Goeddel, D. V., and Ferrara, N. (1989) Vascular endothelial growth factor is a secreted angiogenic mitogen. Science 246, 1306–1309.
  6. Ferrara, N. (1999) Vascular endothelial growth factor: molecular and biological aspects. Curr. Top. Microbiol. Immunol. 237, 1–30.
  7. Ferrara, N. (2000) Vascular endothelial growth factor and the regulation of angiogenesis. Recent Prog. Horm. Res. 55, 15-35-36.
  8. Ishimaru, S., Ueda, R., Hinohara, Y., Ohtani, M., and Hanafusa, H. (2004) PVR plays a critical role via JNK activation in thorax closure during Drosophila metamorphosis. EMBO J. 23, 3984–3994.
  9. Cho, N. K., Keyes, L., Johnson, E., Heller, J., Ryner, L., Karim, F., and Krasnow, M. A. (2002) Developmental control of blood cell migration by the Drosophila VEGF pathway. Cell 108, 865–876.
  10. Sawano, A., Takahashi, T., Yamaguchi, S., and Shibuya, M. (1997) The phosphorylated 1169-tyrosine containing region of flt-1 kinase (VEGFR-1) is a major binding site for PLCgamma. Biochem. Biophys. Res. Commun. 238, 487–491.
  11. Landgren, E., Schiller, P., Cao, Y., and Claesson-Welsh, L. (1998) Placenta growth factor stimulates MAP kinase and mitogenicity but not phospholipase C-gamma and migration of endothelial cells expressing Flt 1. Oncogene 16, 359–367.
  12. Iliopoulos, O., Levy, A. P., Jiang, C., Kaelin, W. G., and Goldberg, M. A. (1996) Negative regulation of hypoxia-inducible genes by the von Hippel-Lindau protein. Proc. Natl. Acad. Sci. U. S. A. 93, 10595–10599.
  13. Reinmuth, N., Fan, F., Liu, W., Parikh, A. A., Stoeltzing, O., Jung, Y. D., Bucana, C. D., Radinsky, R., Gallick, G. E., and Ellis, L. M. (2002) Impact of insulin-like growth factor receptor-I function on angiogenesis, growth, and metastasis of colon cancer. Lab. Investig. J. Tech. Methods Pathol. 82, 1377–1389.
  14. Van Belle, E., Witzenbichler, B., Chen, D., Silver, M., Chang, L., Schwall, R., and Isner, J. M. (1998) Potentiated angiogenic effect of scatter factor/hepatocyte growth factor via induction of vascular endothelial growth factor: the case for paracrine amplification of angiogenesis. Circulation 97, 381–390.
  15. Dong, J., Grunstein, J., Tejada, M., Peale, F., Frantz, G., Liang, W.-C., Bai, W., Yu, L., Kowalski, J., Liang, X., Fuh, G., Gerber, H.-P., and Ferrara, N. (2004) VEGF-null cells require PDGFR alpha signaling-mediated stromal fibroblast recruitment for tumorigenesis. EMBO J. 23, 2800–2810.
  16. Needleman, P., Turk, J., Jakschik, B. A., Morrison, A. R., and Lefkowith, J. B. (1986) Arachidonic acid metabolism. Annu. Rev. Biochem. 55, 69–102.
  17. Fleming, R. Y., Ellis, L. M., Parikh, N. U., Liu, W., Staley, C. A., and Gallick, G. E. (1997) Regulation of vascular endothelial growth factor expression in human colon carcinoma cells by activity of src kinase. Surgery 122, 501–507.
  18. Dias, S., Hattori, K., Heissig, B., Zhu, Z., Wu, Y., Witte, L., Hicklin, D. J., Tateno, M., Bohlen, P., Moore, M. A., and Rafii, S. (2001) Inhibition of both paracrine and autocrine VEGF/ VEGFR-2 signaling pathways is essential to induce long-term remission of xenotransplanted human leukemias. Proc. Natl. Acad. Sci. U. S. A. 98, 10857–10862.
  19. Konishi, T., Huang, C. L., Adachi, M., Taki, T., Inufusa, H., Kodama, K., Kohno, N., and Miyake, M. (2000) The K-ras gene regulates vascular endothelial growth factor gene expression in non-small cell lung cancers. Int. J. Oncol. 16, 501–511.
  20. Fujisawa, T., Watanabe, J., Kamata, Y., Hamano, M., Hata, H., and Kuramoto, H. (2003) Effect of p53 gene transfection on vascular endothelial growth factor expression in endometrial cancer cells. Exp. Mol. Pathol. 74, 276–281.
  21. Zundel, W., Schindler, C., Haas-Kogan, D., Koong, A., Kaper, F., Chen, E., Gottschalk, A. R., Ryan, H. E., Johnson, R. S., Jefferson, A. B., Stokoe, D., and Giaccia, A. J. (2000) Loss of PTEN facilitates HIF-1-mediated gene expression. Genes Dev. 14, 391–396.
  22. Youssoufian, H., Hicklin, D. J., and Rowinsky, E. K. (2007) Review: monoclonal antibodies to the vascular endothelial growth factor receptor-2 in cancer therapy. Clin. Cancer Res. Off. J. Am. Assoc. Cancer Res. 13, 5544s–5548s.
  23. Clin Med Insights Oncol. 2012; 6: 19–30. Published online 2012 Jan 4. doi:  10.4137/CMO.S7432 PMCID: PMC3256978 Aflibercept in the Treatment of Metastatic Colorectal Cancer
  24. Koga, K., Osuga, Y., Yoshino, O., Hirota, Y., Ruimeng, X., Hirata, T., Takeda, S., Yano, T., Tsutsumi, O., and Taketani, Y. (2003) Elevated serum soluble vascular endothelial growth factor receptor 1 (sVEGFR-1) levels in women with preeclampsia. J. Clin. Endocrinol. Metab. 88, 2348–2351.
  25. Hurwitz, H., Fehrenbacher, L., Novotny, W., Cartwright, T., Hainsworth, J., Heim, W., Berlin, J., Baron, A., Griffing, S., Holmgren, E., Ferrara, N., Fyfe, G., Rogers, B., Ross, R., and Kabbinavar, F. (2004) Bevacizumab plus irinotecan, fluorouracil, and leucovorin for metastatic colorectal cancer. N. Engl. J. Med. 350, 2335–2342.
  26. Yang, S., Zhao, J., and Sun, X. (2016) Resistance to anti-VEGF therapy in neovascular age-related macular degeneration: a comprehensive review. Drug Des. Devel. Ther. 10, 1857–1867.
  27. Oosthuyse, B., Moons, L., Storkebaum, E., Beck, H., Nuyens, D., Brusselmans, K., Van Dorpe, J., Hellings, P., Gorselink, M., Heymans, S., Theilmeier, G., Dewerchin, M., Laudenbach, V., Vermylen, P., Raat, H., Acker, T., Vleminckx, V., Van Den Bosch, L., Cashman, N., Fujisawa, H., Drost, M. R., Sciot, R., Bruyninckx, F., Hicklin, D. J., Ince, C., Gressens, P., Lupu, F., Plate, K. H., Robberecht, W., Herbert, J. M., Collen, D., and Carmeliet, P. (2001) Deletion of the hypoxia-response element in the vascular endothelial growth factor promoter causes motor neuron degeneration. Nat. Genet. 28, 131–138.
  28. Verheyen, A., Peeraer, E., Nuydens, R., Dhondt, J., Poesen, K., Pintelon, I., Daniels, A., Timmermans, J.-P., Meert, T., Carmeliet, P., and Lambrechts, D. (2012) Systemic anti-vascular endothelial growth factor therapies induce a painful sensory neuropathy. Brain J. Neurol. 135, 2629–2641.