BOX
By: William Sommer and Daniel Weibel, ChemFiles 2008, 8.2, 64.
ChemFiles 2008, 8.2, 64.
C2-symmetric chiral bisoxazolines (BOX) are privileged structures because they promote a great number of transformations with unprecedented selectivity.1 In 1991, in the same Journal of American Chemistry issue, two communications appeared by Evans2 and Corey.3 These two publications have paved the way for a rapidly and ever-growing number of examples where BOX ligands are used successfully as chiral ligands.
Evans described the cyclopropanation of alkenes using diazo esters catalyzed by a complex formed of 406147 and Cu(I)OTf. With the ester derived from 2,6‑di-tert-butyl-4‑methylphenol (BHT) it was found that the increased steric demand displays increased trans selectivity (Scheme 1).2
Scheme 1
In a related experiment, Evans announced the first catalytic enantioselective aziridination of styrene.4 Two years later, he reported on an extension of this preliminary work in a full communication (Scheme 2). Using BOX ligand 405000, complexed to Cu(I)OTf, in the catalytic enantioselective aziridination of styrene, the BOX ligand having phenyl substituents proved to be superior to the one bearing sterically demanding t-Bu groups (Scheme 2).
Scheme 2
Recently, Reiser et al. discovered a general route to disubstituted γ-butyrolactones. In an Cu(I)-catalyzed asymmetric cyclopropanation of furans with diazo esters, bis(4‑isopropyloxazoline) 680192 was used as chiral ligand (Scheme 3). This transformation was carried out on 50–100 g scale without a detectable loss of enantioselectivity.5
Scheme 3
In his 1991 Journal of American Chemical Society communication, Corey used BOX ligand 405000 in an Fe(III)-catalyzed enantioselective Diels–Alder reaction (Scheme 4).3
Scheme 4
Two year later, Evans described the Cu(II)-catalyzed version of the enantioselective Diels–Alder reaction of unsubstituted acrylimides using BOX ligand 406147 as a chiral Lewis acid.6 The reaction proceeded in good yield exhibiting excellent enantioselectivity for the endo diastereoisomer (Scheme 5). From further investigations by Evans et al., it appears that the best combination is BOX 406147 and Cu(II), and the best counterions are OTf and SbF6.1
Scheme 5
4‑Aryl- and 4‑alkylsubstituted BOX-based catalysts were widely used in a large number of reactions1and their behavior can provide a good representation of their efficiency. But BOX ligands with other structural motifs have been successfully used too. BOX ligand 405981 has been used in the Cu(I)-catalyzed highly enantioselective Mannich reactions to efficiently produce highly functionalized 4‑oxo-glutamic ester derivatives (Scheme 6).7
Scheme 6
Recently, Ma et al. reported on the enantioselective CuI/bis(oxazoline)- catalyzed addition of propiolates and terminal ynones to 1‑acylpyridinium salt to afford highly functionalized dihydropyridines with excellent enantioselectivity using 464155 (Scheme 7).8 It is found that the carbonyl group adjacent to the alkyne moiety is essential for the enantioselectivity of the addition.
Scheme 7
back to top
Materials
References
  1. (a) Desimoni, G.; Faita, G.; Jørgensen, K. A. Chem. Rev. 2006, 106, 3561‑3651. (b) Ghosh, A. K.; Mathivanan, P., Cappiello, J. Tetrahedron: Asymmetry 1998, 9, 1‑45. (c) Johnson, J. S.; Evans, D. A. Acc. Chem. Res. 2000, 33, 325‑335.
  2. Evans, D. A.; Woerpel, K. A.; Hinman, M. M.; Faul, M. M. J. Am. Chem. Soc. 1991, 113, 726.
  3. Corey E. J.; Imai, N.; Zhang, H.-Y. J. Am. Chem. Soc. 1991, 113, 728.
  4. Evans, D. A.; Faul, M. M.; Bilodeau, M. T.; Anderson, B. A.; Barnes, D. M. J. Am. Chem. Soc. 1993, 115, 5328‑5329.
  5. (a) Jezek, E.; Schall, A.; Kreitmeier, P.; Reiser, O. Synlett 2005, 915‑918. (b) Chhor, R. B.; Nosse, B.; Sörgel, S.; Böhm, C.; Seitz, M.; Reiser, O. Chem. -Eur. J. 2003, 9, 260‑270.
  6. Evans, D. A.; Miller, S. J.; Lecta, T. J. Am. Chem. Soc. 1993, 115, 6460‑6461.
  7. Juhl, K.; Gathergood, N.; Jørgensen, K. A. Angew. Chem., Int. Ed. 2001, 40, 2995.
  8. Sun, Z.; Yu, S.; Ding, Z; Ma, D. J. Am. Chem. Soc. 2007, 129, 9300‑9301.