Chemfiles Volume 7 Article 5
Until recently, mild aromatic C–O bond formation was exceedingly difficult to achieve. Palladium complexes of Q-Phos catalyze the mild etherification reaction of aryl halides with alkoxide, siloxide, and phenoxide partners (Table 2).1,2 Reactions with aryl bromides, or electron-poor aryl chlorides, typically proceed at room temperature. The catalyst system was also successful in the synthesis of coumarans, via intramolecular etherification.
Table 2.
back to top
  1. Kataoka, N. et al. J. Org. Chem. 2002, 67, 5553.
  2. Shelby, Q. et al. J. Am. Chem. Soc. 2000, 122, 10718.