Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Highly Acid-Labile Resins for the Synthesis of Protected Peptide Carboxamides
ChemFiles Volume 3 Article 4
This extremely acid-labile resin provides Fmoc-/tBu-protected peptide amides by treatment with low concentrations of TFA (~ 1%).(1-2) Reductive alkylation of this support followed by acylation gives access to secondary carboxamide products.(3)
back to top 
Materials
References
  1. Albericio, F.; Barany, G. Int. J. Pept. Protein Res. 1987, 30, 206.
  2. Albericio, F. et al. J. Org. Chem. 1990, 55, 3730.
  3. Songster, M. F. et al. Lett. Pept. Sci. 1995, 2, 265.