Better Data Handling with 2D Smart Labels
 No typos, no manually writing, no outdated safety data!
With our new smart label with 2D Data Matrix barcode, you're just one touch away from digitalized and up-to-date product data.
Smart labels contain a 2D data matrix barcode. The 2D Data Matrix barcode contains the product data encoded using Global Standard One (GS1) specifications, which allows for processing directly in your LIMS or ERP system.This barcode provides seamless access to essential product information (item code, batch number, shelf life, etc.) and product documentation (CoA, SDS, etc.).
Benefits
  • Easy, quick and convenient access to product data
  • Seamless access to safety data
  • Mobile access via App
  • Simple access via website with a hand-held scanner
  • Direct processing in LIMS or ERP
 3 smart ways to easy data access – choose what works for you
My M Safety App
Use your smartphone and our new My M Safety app.
Video
Apple App Store
Google Play Store
Scan Now Website
Use a common hand held scanner connected to your PC or laptop. Scan the 2D barcode out of your LIMS or ERP system to directly insert relevant product data in your application.
Scan Now Website
Barcode Scanner
Scan the 2D barcode out of your LIMS or ERP system to directly insert relevant product data in your application.
Video