Head HTML-
Carp Kidney
Submitted by: Mr Stanislav Antonov
Institute of Molecular Genetics of RAS
This is a carp (cyprinus carpio) kidney paraffin section (6 micron) produced with a Brunel rocking microtome. Image reveals proximal (dark red) and distal (pink with blue outlines) convoluted tubules as well as the surrounding hematopoietic tissue. Image was taken with Amscope T340B
Magnification: 400x
Back to Gallery