Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Protein Of The Month
February: β-Glucuronidase
Origin: β-Glucuronidase is a family of enzymes that perform a diverse set of functions. In humans, for example, it converts conjugated bilirubin into the unconjugated form, making bilirubin suitable for reabsorption.
Application: β-Glucuronidase is used in laboratories as a reporter gene in GUS assays to monitor gene promotor activity.
How: β-Glucuronidase catalyzes the breakdown of complex carbohydrates. The GUS assay combines β-Glucuronidase with a colourless or non-fluorescent substrate. An active promoter may then induce expression of β-Glucuronidase, which in turn then transforms the substrate to a coloured or fluorescent product.
Did you know: Although these associations are modest, some studies suggest that plant foods, particularly constituents of fruits and fiber-containing foods, may impact human β-glucuronidase activity in what can be viewed as a potentially favorable direction.
Learn More:
Return to mainpage