Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Protein of the month
Protein of the Month, May: Gelatin from porcine skin (G2500)
General Description: Gelatin is a mixture of peptides and proteins produced by partial hydrolysis of collagen extracted from skin and bones.
Application: Gelatin is used for coating cell culture plates to improve cell attachment for a variety of cell types, used in PCR to help stabilize Taq DNA polymerase and use as a blocking reagent in western blotting, ELISA and immunohistochemistry. Gelatin also has many industrial applications such as a stabilizer, thickener, and texturizer.
Did you know? In 1754, the first English patent for the manufacturing of gelatin was granted. Unflavored dried gelatin became available in 1842 from the J and G Company in Edinburgh, Scotland.
  Learn more:
Return to mainpage