Product for QuEChERS Method for matrices containing > 15% fat:
Supel™ QuE Z-Sep+
Save 20%
Please mention Promotion Code HGK when ordering, valid until October 31, 2017*
Supel™ QuE Z-Sep+ enhances sample cleanup for complex matrices by effectively removing more fat and color from sample extracts than traditional phases for QuEChERS LC-MS methods. By eliminating problematic matrix interferences, Z-Sep products provide more robust LC-MS methods. This proprietary technology can replace C18 and PSA in your current methods without additional method development. Z-Sep+ is recommended for cleanup of samples containing greater than 15% fat.
Product No. Description Discount [%] Add to Cart
2mL
55172-U PK50 SUPEL QUE PSA (EN) 20
55173-U PK50 SUPEL QUE PSA/C18 (EN) 20
55147-U PK50 SUPEL QUE PSA/ENVI-CARB (EN) TUBE 1 20
55176-U PK50 SUPEL QUE PSA/ENVI-CARB (EN) TUBE 2 20
55287-U PK50 SUPEL QUE PSA (AOAC) TUBE 2 20
55288-U PK50 SUPEL QUE PSA/C18 (AOAC) TUBE 2 20
55289-U PK50 SUPEL QUE PSA/C18/ENVI-CARB (AOAC) 20
12mL
55228-U PK50 SUPEL QUE PSA (EN) 20
55229-U PK50 SUPEL QUE PSA/C18 (EN) 20
55230-U PK50 SUPEL QUE PSA/ENVI-CARB (EN) TUBE 1 20
55233-U PK50 SUPEL QUE PSA/ENVI-CARB (EN) TUBE 2 20
55282-U PK50 SUPEL QUE PSA (AOAC) TUBE 2 20
55283-U PK50 SUPEL QUE PSA/C18 (AOAC) TUBE 2 20
55286-U PK50 SUPEL QUE PSA/C18/ENVI-CARB (AOAC) 20
15mL
55437-U PK50 SUPEL QUE PSA (EN) 20
55439-U PK50 SUPEL QUE PSA/C18 (EN) 20
55446-U PK50 SUPEL QUE PSA/ENVI-CARB (EN) TUBE 1 20
55464-U PK50 SUPEL QUE PSA/ENVI-CARB (EN) TUBE 2 20
55466-U PK50 SUPEL QUE PSA (AOAC) TUBE 2 20
55470-U PK50 SUPEL QUE PSA/C18 (AOAC) TUBE 2 20
55474-U PK50 SUPEL QUE PSA/C18/ENVI-CARB (AOAC) 20
Products for the extraction step
55227-U SUPEL QUE Citrate (EN) 20
55237-U SUPEL QUE Citrate/Sodium Bicarbonate (EN) 20
Back to Top
Supel is a trademark of Sigma-Aldrich Co. LLC
*Offers valid in EMEA throughout the validity period stated. Certain restrictions apply. Offers do not apply to products sold through Sigma-Aldrich dealers or to customers with contracts in place. Certain offers may not be valid in all geographic regions; additional restrictions may apply - please contact your local office for information specific to your region. Sigma-Aldrich reserves the right to modify or terminate these or any other promotions at any time in its sole discretion.