Process Development Services
Fast Forward to Cell Therapy Success
On the cutting edge of regenerative medicine, you need a partner with a proven track record of advancing cellular therapy product development and manufacturing, and the ability to adapt to your needs.
As your trusted expert in Life Science, we provide optimized cell therapy manufacturing platforms, media and reagents, characterization tools, and testing – all required to tackle the bottlenecks of cell therapy commercialization. Our Cell Therapy Process Development Services allow you to:
  • Accelerate successful research into scalable industrialized processes
  • cGMP-compatible
  • Increase patient access through cost-effective, robust manufacturing
  • Remove subjectivity and operator error through closed and automated processes
  • Prepare yourself (or your CMO) to advance quickly, based on our technical expertise and knowledge transfer
Allogeneic Cell Therapy: Large-scale manufacturing; 1 donor = multiple patient doses
Featured service highlight
Robust stirred tank processes allow for successful industrialized manufacturing with an average of 34% cost reduction over planar processes.
Production Costs, Normalized
Planar
50L
Materials and reagents Cost, Normalized
1.00
0.80
Volume media (L)
142
93
Production vessels
55
1
Labor
1.00
0.49
Max parallel operators
11
2
Total labor hours
173
87
Total Cost per Dose (100M Cells)
1.0
0.66
Materials and Reagents % of Total Cost
80%
88%
Labor % of Total Cost
20%
12%
*Values based on 25 billion target yield. Cost of medium in bulk = $200 / L
To learn more about our Process Development Services for Cell Therapy Manufacturing, contact us today.
Contact us
We are devoted to meeting all your cell therapy manufacturing needs. Our extensive stem cell expertise and manufacturing experience make us an ideal partner to advance and translate your work to cell therapy success. Connect with us today to discuss your specific application needs.