[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Pathogen & Spoilage Testing](https://www.sigmaaldrich.com/US/en/applications/microbiological-testing/pathogen-and-spoilage-testing)Microbiological Control of Spices and Herbs
# Microbiological Control of Spices and Herbs
__Jvo Siegrist__
Cultures around the world rely on herbs and spices to add flavor and zest to food. Many spices, however, contain very high numbers of bacteria, making them a potent source for food spoilage and pathogens.
To study the microbiological status of herbs and spices, E. de Boer et al.1 tested 150 samples collected from 54 different spices, spice mixtures and herbs. They reported at least 1,000 organisms per gram, with most spices containing 105-106 cells per gram. A high number of psychotropic bacteria, yeasts and Enterobacteriaceae was detected mainly on herbal spices originating in moderate climate areas. The study also reported high mold counts, identifying *Aspergillus niger, A. flavus, A. tamarii, Penicillium citrinium, P. chrysogenum, and Absidia corymbifera* as the most frequent isolated species. Since A. flavus may produce aflatoxins, one of the most potent naturally occurring toxins, its presence should be a matter of concern and monitored closely by the spice industry. Another serious potential public health risk may involve the presence of pathogenic bacteria; frequently reported species are *Clostridium perfringens, Bacillus cereus and Salmonella*.
Several issues present a challenge in the microbiological study of dried herbs and spices. Dryness, inhibiting substances, high osmotic pressure, and other adverse conditions heavily stress the cells. Long stress periods can eventually lead the cells to a “viable but not culturable“ (VNC) state. In a VNC state, the microbes cannot grow on conventional laboratory plating media but may revive in vivo and cause disease. However, with the addition of certain growth factors, traditional media can be upgraded and VNC organisms can be resuscitated.
One of these growth factors is ferrioxamine E, which significantly improves the recovery of Salmonella, Cronobacter spp., Staphylococcus aureus and Yersinia enterocolitica from contaminated foods2-4. A concentration of ferrioxamine E in the range of 5-200 ng/ mL supports reproducible growth (__Table 1__). Ferrioxamine E provides the essential bio-available form of micronutrient iron (III) to the organisms. This leads to a reduced lag-phase in the medium and reactivates damaged bacteria, leading to optimal microbial analyses.
__Figure 1.__Spices of plant origin
__Figure 2.__Structure of ferrioxamine E
| | |
|---------------------------------------------|-------|
| Organisms | ng/mL |
| *Salmonella* | 75 |
| *Cronobacter spp. (Enterobacter sakazakii)* | 150 |
| *Yersinia enterocolitica* | 100 |
Table 1.Recommended end concentration of ferrioxamine E
__Figure 3.__HiCrome™ Bacillus Agar
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### References
1\.
de Boer E, Spiegelenberg WM, Janssen FW. 1985. Microbiology of spices and herbs. Antonie van Leeuwenhoek. 51(4):435-438. [https://doi.org/10.1007/bf02275058](https://doi.org/10.1007/bf02275058)
2\.
Makino S, Kii T, Asakura H, Shirahata T, Ikeda T, Takeshi K, Itoh K. 2000. Does Enterohemorrhagic Escherichia coliO157:H7 Enter the Viable but Nonculturable State in Salted Salmon Roe?. Appl. Environ. Microbiol.. 66(12):5536-5539. [https://doi.org/10.1128/aem.66.12.5536-5539.2000](https://doi.org/10.1128/aem.66.12.5536-5539.2000)
3\.
Barcina I, Lebaron P, Vives-Rego J. Survival of allochthonous bacteria in aquatic systems: a biological approach. 23(1):1-9. [https://doi.org/10.1111/j.1574-6941.1997.tb00385.x](https://doi.org/10.1111/j.1574-6941.1997.tb00385.x)
4\.
Cho J, Kim S. 1999. Viable, but non-culturable, state of a green fluorescence protein-tagged environmental isolate ofSalmonella typhiin groundwater and pond water. 170(1):257-264. [https://doi.org/10.1111/j.1574-6968.1999.tb13382.x](https://doi.org/10.1111/j.1574-6968.1999.tb13382.x)
__Related Articles__
- [*Lactobacilli* Basics, Testing & Identification](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/lactobacilli)
- [Microbial Detection Using Chromogenic and Fluorogenic Culture Media](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/overview-chromogenic-media)
- [Identification of Microorganisms Based on Color](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/cytology-and-microscopy/color-identification-microorganisms)
- [Culture Method for Beer Quality Control](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/culture-method-beer-quality)
- [Detection and Differentiation of Staphylococcus Aureus on Baird Parker Agar](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/detection-and-differentiation)
- [Differentiation of Escherichia coli from coliforms](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/differentiation-of-e-coli-coliforms)
- [DiluCult™ Gravimetric Dilutor & Sample Homogenizer](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/dilucult-gravimetric-dilutor-homogenizer)
- [EN ISO 11133 Microbiological Culture Media](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/en-iso-11133-water-for-the-preparation-and-performance-testing)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Molecular Cloning & Protein Expression](https://www.sigmaaldrich.com/US/en/products/molecular-biology-and-functional-genomics/cloning-and-expression/molecular-cloning-and-protein-expression)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
An unknown error has occured.