医薬品中不純物
医薬品不純物 混合標準溶液
医薬品不純物として指定されているRelatedCompound(USP)、Impurity(EP)の物質を溶媒で希釈し、単一および混合溶液の、認証標準物質(CRM)でご提供します。
【特長】
  • 信頼性確保に不可欠な認証標準物質(CRM)
  • 溶液の為、ハンドリングしやすく、人による作業中のエラーが起こりにくい
  • 混合標準溶液として調整しているためそれぞれの物質を買わなくて済み、コストを削減できる
Cat. No. 製品名 成分 濃度 (µg/mL) 品質グレード
A-134 Acetaminophen (Paracetamol) Impurity Mixture API, Related Compound B, Related Compound C, Related Compound D, Related Compound F 25 CRM
A-135 Acetaminophen Related Compound A (4'-Acetoxyacetanilide) Related Compound A 250 CRM
A-136 Acetaminophen Related Compound K (4-Aminophenol) Related Compound K 250 CRM
C-166 Acetaminophen Related Compound J (4-Chloroacetanilide) Related Compound J 250 CRM
F-052 Furosemide Impurity Mixture API, Related Compound A, Related Compound B 5 CRM
G-022 Guaifenesin Impurity Mixture API, Impurity A, Impurity B 500-1000 CRM
I-036 Ibuprofen Impurity Mixture API, Impurity A, Related Compound C, Related Compound J, Impurity N 25 CRM
I-037 Ibuprofen Impurity B ((2RS)-2-(4-butylphenyl) propanoic acid) Impurity B 100 CRM
L-048 Lidocaine Impurity Mixture API, Impurity A, Impurity H 20-200 CRM
M-195 Metformin Impurity Mixture API, Related Compound B, Related Compound C, Impurity D 50 CRM
M-196 Metformin Related Compound A (Cyanoguanidine) Related Compound A 50 CRM
O-048 Omeprazole Impurity Mixture API, Related Compound A, Related Compound B, Related Compound E, Related Compound I 60 CRM
P-125 Parabens Impurity Mixture Methyl, Ethyl, Propyl, Butyl Parabens 50 CRM
P-126 Pyridoxine Impurity Mixture API, Impurity A, Impurity B 50 CRM
S-100 Salicylic Acid Impurity Mixture API, Related Compound A, Related Compound B, Impurity C 10-500 CRM