Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
カスタム(シグマジェノシス)製品 | カスタムペプチド合成 | Custom Peptide Synthesis Service
Sigma-Aldrich can provide high-quality synthetic peptides. Peptides are delivered with MALDI-TOF/MS and HPLC data. Additionally, general guideline of peptide handling and storage is attached.
Please refer to this guideline on dissolving. And the check sheet is also attached, which says the amino acid composition of your peptide sequences and isoelectric point.
Specification
Purity Yield shipping schedule
Immunological Purity
(Crude)
5mg 3 weeks
10mg
15mg
25mg
>70% Purity 5mg 3 weeks
10mg
15mg
25mg
>80% Purity 5mg 3-4 weeks
10mg
15mg
25mg
>90% Purity 5mg 4-5 weeks
10mg
15mg
25mg
>95% Purity 5mg 4-5 weeks
10mg
15mg
25mg
*Please inquire about price of peptide
*Shipping schedule in the table, for the peptide sequence length below 21AA. Please inquire about over 21AA peptide.
Terminal modification / Modified amino acids available
Fluorescein pY N-terminal Acetylation
Biotin pS C-terminal Amidation
Myristoyl pT KLH/BSA carrier protein conjugation
*Please inquire about price and shipping schedule of modified peptide.
Proven Terminal modification / Modified amino acids
C-terminal NH2 of side chain Modified amino acid
Amidation Biotin 3-Nitro-Tyrosine
  Fluorescein 6-Aminocaproic acid(Ahx)
N-terminal   Aminobutyric acid (Abu)
Acetylation  SH of side chain Citrulline
Biotin Biotin Cysteine(Acm protected) 
Boc(butoxycarbonyl)   D-Amino acid 
Dabcyl Atto488 Dimethy-Lysine
Dabsyl Atto550 Hydroxy-Proline
DHA Atto633 Methyl-Lysine
Formylation Atto647N Norleucine(Nle)
Myristoylation Atto655 Ornithine
Nicotinic acid  Dansyl Phosphorylation
Palmitylation Fluorescein Pyroglutamic acid 
Z.Cbs(Carbobenzoxyl) RG(Rhodamine Green)  Trimethyl-Lysine
  Rhodamine B β-Alanine
special treatment Tamra(Tetramethylrhodamine) ε-Acetyl-Lysine
Branching Texas Red   
Cyclization    
Carrier protein conjugation   etc.
For handling and storage of peptides
Please see Synthetic Peptide Storage and Handling guide (PDF: 458kb).
DNA Calculatorが新機能を追加してリニューアル!
【主な機能】
  • Analysis:分子量、Tm値、ダイマー、ヘアピン予測等。DNA/RNA/特殊塩基にも対応します。
  • Resuspension:核酸の溶解液量の計算をサポートします。
  • Dilution:核酸溶液の希釈計算をサポートします。
※オリゴ購入の会員登録とは別にユーザー登録が必要となります。
大変お手数ですが上記リンク先からご登録の上、ご利用ください。
 Antibody Search Tool
Help Perform full text search of antibodies and related products using keywords. "Refine Your Search" and search Within Results options are available from the search results page.
For Primary Antibodies
Enter specificity, gene symbol, gene ID, or gene family name (use "*") and other relevant keywords: "actin", "mouse monoclonal actin", "actin Cy3", "rabbit actin", "XRCC1", "22594", "XRCC*". Click the "GO" button.
For Secondary Antibodies
Enter IgG target, label, and host species: "rabbit IgG FITC mouse", "mouse IgG TRITC", "goat IgG peroxidase". Click the "GO" button.
Refine search if necessary using "Secondary Antibodies" option from "Refine Your Search" under "Product Category".
For a Clone Search
Enter Clone name or number:"1A4", "V9". Click the "GO" button.