タンパク質検出と測定・定量
ELISA
Sigma-Aldrichはpgレベルで検出可能な96ウェルプレートにプレコーティングされたELISAキットを多数ご用意しています。
∇ Sigma-AldrichのELISAキットの検索はこちら
∇ メルク(旧Chemicon, Linco Research)製品はこちら
神経研究用 ELISAキット
測定項目 動物種 スタンダード範囲 感度 サンプル量 カタログ番号
Amyloid beta 1-40 Human 16–500 pg/mL 4 pg/mL 50 μL EZHS40
Amyloid beta 1-42 Human 16–500 pg/mL 5 pg/mL 50 μL EZHS42
Amyloid beta, Set Human EZHS40 と EZHS42 各1キットずつを含むセット EZHS-SET
Amyloid beta (Brain) 1-40 Human 16–500 pg/mL 4 pg/mL 50 μL EZBRAIN40
Amyloid beta (Brain) 1-42 Human 16–500 pg/mL 5 pg/mL 50 μL EZBRAIN42
Amyloid beta (Brain), Set Human EZBRAIN40 と EZBRAIN42 各1キットずつを含むセット EZBRAIN-SET
BDNF (Brain-Derived Neurotrophic Factor) Human, Rat 7.8–500 pg/mL 7.8 pg/mL 50 μL CYT306
GFAP (Glial Fibrillary Acidic Protein) Human, Mouse, Rat 1.5-100 ng/mL 1.5 ng/mL 100 μL NS830
NGF (Nerve Growth Factor) Mouse, Rat 10–1000 pg/mL 10-15 pg/mL 50 μL CYT304
NPY (Neuropeptide Y) Human 5-1000 pg/mL 2 pg/mL 50 μL EZHNPY-25K
NPY (Neuropeptide Y) Mouse, Rat 0.01-2 ng/mL 0.004 ng/mL 20 μL EZRMNPY-27K
PEDF (Pigment Epithelium-Derived Factor) Human 0.9–62.5 ng/mL 0.9 ng/mL 50 μL CYT420
Phosphorylated Neurofilament, (pNF-H) Sandwich Multi-Species 0.0293–15 ng/mL 0.0585 ng/mL 1-10 μL NS170
S100B Human 2.7-2000 pg/mL 1.3 pg/mL 50 μL EZHS100B-33K
代謝研究用 ELISAキット
測定項目 動物種 スタンダード範囲 感度 サンプル量 カタログ番号
Adiponectin Human 1.56–200 ng/mL 0.2 ng/mL 10 μL EZHADP-61K
Adiponectin (High Molecular Weight) Human 1.5–200 ng/mL 0.5 ng/mL 10 μL EZHMWAN-65K
Adiponectin Mouse 1–50 ng/mL 0.2 ng/mL 10 μL EZMADP-60K
Adiponectin Rat 3.125–200 ng/mL 0.4 ng/mL 11 μL EZRADP-62K
Amylin (active) Human 1–100 pM 0.7 pM 50 μL EZHA-52K
C-Peptide Human 0.2–20 ng/mL 0.05 ng/mL 10 μL EZHCP-20K
C-Peptide Canine 0.2–10 ng/mL 0.24 ng/mL 25 μL EZCCP-47K
C-Peptide 2 Mouse, Rat 25–1600 pM 15.0 pM 20 μL EZRMCP2-21K
FGF-21 Human 31.25–2000 pg/mL 10.0 pg/mL 50 μL EZHFGF21-19K
FGF-21 Mouse, Rat 49.4–12,000 pg/mL 10.0 pg/mL 10 μL EZRMFGF21-26K
FGF-23 Human 9.9–2400 pg/mL 3.5 pg/mL 50 μL EZHFGF23-32K
GIP (total) Human 8.2–2000 pg/mL 4.2 pg/mL 20 μL EZHGIP-54K
GIP (total) Mouse, Rat 8.2–2000 pg/mL 4.2 pg/mL 10 μL EZRMGIP-55K
GLP-1 (active) Multi-Species 2–100 pM 1.0 pM 100 μL EGLP-35K
GLP-1 High Sensitivity (active)  (化学発光) Multi-Species  See data sheet 0.14 pM 50 μL EZGLPHS-35K
GLP-1 (total) Multi-Species 4.1–1000 pM 1.0 pM 20-50 μL EZGLP1T-36K
GLP-2 Multi-Species 1–64 ng/mL 0.3 ng/mL 50 μL EZGLP2-37K
Glucagon (化学発光)  Human,Mouse, Rat 0.02–2 ng/mL 0.003 ng/mL 150-300 μL EZGLU-30K
Growth Hormone (GH) Mouse, Rat 0.7–50 ng/mL 0.07 ng/mL 10 μL EZRMGH-45K
Insulin WHO Human 2–200 μU/mL 1.0 μU/mL 20 μL EZHI-14K
Insulin Mouse, Rat 0.2–10 ng/mL 0.1 ng/mL 10 μL EZRMI-13K
Insulin (Animal serum free) Human 2–200 μU/mL 0.85 μU/mL 20 μL EZHIASF-14K
Leptin Canine 0.78–50 ng/mL 0.21 ng/mL 20 μL EZCL-31K
Leptin Mouse 0.2–30 ng/mL 0.05 ng/mL 10 μL EZML-82K
Leptin Rat 0.2–30 ng/mL 0.08 ng/mL 10 μL EZRL-83K
Leptin “Dual Range” WHO Human 0.5–100 ng/mL 0.2 ng/mL 25 μL EZHL-80SK
Procollagen Type IIA N-Propeptide (PIIANP) Human lot dependent 30.0 ng/mL 5 μL EZPIIANP-53K
Proinsulin (total) WHO Human 2–200 pM 0.5 pM 20 μL EZHPI-15K
PYY (total) Human 10–2000 pg/mL 6.5 pg/mL 20 μL EZHPYYT66K
Resistin Human 0.16–5 ng/mL 0.02 ng/mL 20 μL EZHR-95K
SAA-3 Mouse 0.078-5 μg/mL 0.078 μg/mL 10 μL EZMSAA3-12K