細胞アプリケーション
【参考】他社酵素対応表
メーカー別 酵素対応表
研究の用途に適したリベラーゼ酵素を見つけるために、以下のメーカ別製品リストを参照ください。
酵素を選択したら、アプリケーションにおける結果を向上させるために、現在お使いのコラゲナーゼ濃度を、リベラーゼ リサーチグレードにおける酵素濃度(WU/ml)に変換してください。
実際の作業濃度については「分散酵素の最適化」ページの「酵素の作業濃度」を参照してください。
現在使用しているコラゲナーゼのメーカはどれですか?
> Sigma
> Worthington
> Serva
> Gibco
> Roche
Sigma
Type Specification Wünsch Units/mg Liberase Research Grade
I Crude 0.16 Liberase TM Research Grade
I Sterile 0.16 Liberase TM Research Grade
II Crude 0.32 Liberase DH Research Grade
II Sterile 0.32 Liberase DH Research Grade
III Purified 0.40 Liberase TM Research Grade
IV Crude 0.32 Liberase TM Research Grade
IV Sterile 0.32 Liberase TM Research Grade
V Crude 0.51 Liberase TL Research Grade
V Sterile 0.51 Liberase TL Research Grade
VII Purified, culture tested 2.00 Liberase TM Research Grade
VII Purified 2.00 Liberase TM Research Grade
VII Purified,sterile 2.00 Liberase TM Research Grade
VII Purified, sterile, culture tested 2.00 Liberase TM Research Grade
VIII Crude 0.32 Liberase TM Research Grade
XI Crude 0.90 Liberase TL Research Grade
XI Culture tested 0.90 Liberase TL Research Grade
XI Sterile 0.90 Liberase TL Research Grade
XI Sterile, culture tested 0.90 Liberase TL Research Grade
IA Crude 0.32 Liberase TM Research Grade
IA Culture tested 0.32 Liberase TM Research Grade
IA Sterile 0.32 Liberase TM Research Grade
IA Sterile, culture tested 0.32 Liberase TM Research Grade
F Blend 0.51 Liberase DH Research Grade
H Blend 0.33 Liberase TM Research Grade
L Blend 0.18 Liberase TM Research Grade
N Blend 0.03 Liberase TH Research Grade
Worthington
Type Specification Wünsch Units/mg Liberase Research Grade
1 Crude 0.125 Liberase TM Research Grade
1 Filtered 0.125 Liberase TM Research Grade
2 Crude 0.125 Liberase TH Research Grade
2 Filtered 0.125 Liberase TH Research Grade
3 Crude 0.10 Liberase TM Research Grade
3 Filtered 0.10 Liberase TM Research Grade
4 Crude 0.16 Liberase DH Research Grade
4 Filtered 0.16 Liberase DH Research Grade
Collagenase Purified 0.30 Liberase TM Research Grade
Serva
Type Specification Wünsch Units/mg Liberase Research Grade
Coll. Hep + Crude 0.70 Liberase TM Research Grade
Coll. Pan + Crude 0.70 Liberase TL and DL Research Grade
Collagenase Crude 0.70 Liberase TM Research Grade
Gibco
Type Specification Wünsch Units/mg Liberase Research Grade
Hepatocyte qualified Crude 0.20 Liberase TM Research Grade
I Crude 0.20 Liberase TH Research Grade
II Crude 0.20 Liberase TM Research Grade
III Crude 0.20 Liberase TM Research Grade
IV Crude 0.20 Liberase TL and DL Research Grade
Roche Traditional Collagenase
Type Specification Wünsch Units/mg Liberase Research Grade
A Crude 0.15 Liberase TM Research Grade
B Crude 0.15 Liberase TH Research Grade
D Crude 0.15 Liberase TM Research Grade
P Crude 1.50 Liberase TL and DL Research Grade
H Crude 0.15 Liberase TM Research Grade
ゲノミクス、プロテオミクス、細胞アプリケーション用の研究用試薬・キットがSigma-Aldrichからご利用いただけます