有機エレクトロニクス材料
透明導電性基板 (ITO・FTO)
FTO透明導電性ガラス
FTO(Fluorine-doped tin oxide)透明導電性ガラスは、オプトエレクトロニクスやタッチスクリーン、薄膜太陽電池、省エネ窓、RFI/EMI遮蔽をはじめとするさまざまな用途に理想的な材料です。フッ素ドープ酸化スズは、大気条件下で比較的安定であり、化学的に安定で機械強度が高く、さらに、高温耐熱性をもち、物理的剥離にも強く、酸化インジウムスズよりも安価なため、非常に有望な材料です。
TEC Glass™は、コストパフォーマンスに優れていることで知られています。さらに、FTOを用いた基板は、TiO2やZnOナノ構造薄膜へのインジウムの拡散の問題を避けることができ、熱特性や光透過性に優れています。
製品名 サイズ(L x W x D) 透過率(%) 表面抵抗(Ω/sq) 製品番号
Fluorine doped tin oxide coated glass slide, FTO, TEC 7 50 × 50 × 2.2 mm 80-82 7 735140-5EA
100 × 100 × 2.3 mm 80-82 7 735159-5EA
300 × 300 × 2.2 mm 80-82 7 735167-1EA
Fluorine doped tin oxide coated glass slide, FTO, TEC 8 50 × 50 × 3 mm 80-82 8 735175-5EA
100 × 100 × 3 mm 80-82 8 735183-5EA
300 × 300 × 3.2 mm 80-82 8 735191-1EA
Fluorine doped tin oxide coated glass slide, FTO, TEC 10 50 × 50 × 3 mm 83 10 735205-5EA
100 × 100 × 3 mm 83 10 735213-5EA
300 × 300 × 3 mm 83 10 735221-1EA
Fluorine doped tin oxide coated glass slide, FTO, TEC 15 50 × 50 × 2.2 mm 82-85 13 735248-5EA
100 × 100 × 2 mm 82-85 13 735256-5EA
300 × 300 × 2.3 mm 82-85 13 735264-1EA
* “TEC” is a trademark owned by Nippon Sheet Glass Co. Ltd, or a subsidiary thereof.
ITO透明導電性基板
ITO基板外観
プラスチック基板
製品名 サイズ(L x W x D) ITO膜厚 (Å) 表面抵抗(Ω/sq) 透過率(%) 製品番号
Indium oxide coated PET, IO-PET, Slide 150 x 150 x 0.2 mm - ≦ 10 - 700177-5PAK, 10PAK
Indium tin oxide coated PET, ITO-PET, Sheet 1 1 ft x 1 ft x 5 mil 1,000 60 >79 639303-1EA, 5EA ※1
750 100 >77 639281-1EA, 5EA ※1
1 ft x 1 ft x 7 mil 1,000 60 - 749729-1EA, 5EA
1 ft x 1 ft x 7 mil 1,000 100 - 749737-1EA, 5EA
1 ft x 1 ft x 5 mil 1,000 200 - 749745-1EA, 5EA
1 ft x 1 ft x 7 mil 1,000 200 - 749753-1EA, 5EA
1 ft x 1 ft x 5 mil 1,000 250 - 749761-1EA, 5EA
1 ft x 1 ft x 7 mil 1,000 250 - 749788-1EA, 5EA
1 ft x 1 ft x 5 mil 1,000 300 - 749796-1EA, 5EA
1 ft x 1 ft x 7 mil 1,000 300 - 749818-1EA, 5EA
* 1 mil = 1/1000 inch = 25.4 μm (5 mil = 0.127 mm, 7 mil = 0.178 mm)
※1 A radius of curvature of up to 75 mm will not damage the ITO coating. The polyester film products comprise sputtered ITO, and exhibit transmittance and resistance characteristics similar to those of the glass substrates.
ガラス基板
製品名 サイズ(L x W x D) ITO膜厚 (Å) 表面抵抗(Ω/sq) 透過率(%) 製品番号
Indium tin oxide coated glass slide, rectangular, ITO 1 75 x 25 x 1.1 mm 1,200-1,600 8 – 12 >83 578274-10PAK, 25PAK
600-1,000 15 – 25 >78 636916-10PAK, 25PAK
300-600 30 – 60 >84 636908-10PAK, 25PAK
150-300 70 – 100 >87 576352-10PAK, 25PAK
Indium tin oxide coated glass slide, square, ITO 2 25 x 25 x 1.1 mm 1,200-1,600 8 – 12 ~84 703192-10PAK
300-600 30 – 60 ~84 703184-10PAK
150-300 70 – 100 ~86 703176-10PAK
Indium tin oxide coated aluminosilicate glass slide 3 75 x 25 x 1.1 mm 1,200-1,600 5 – 15 >85 576360-10PAK, 25PAK
1. The slide is made with soda lime float glass. Resistance may increase to as high as 375 Ω when exposed to temperatures of 300 °C for 30 minutes or more. ITO coated float glass is only offered with a SiO2 passivation layer applied directly on the glass prior to ITO coating. 2. Substrate: unpolished float glass, SiO2 passivated (200-300 Å) 3. No passivation layer is present. ITO is directly applied to the substrate. Typical composition: 55.0% SiO2, 7.0% B2O3, 10.4% Al2O3, 21.0% CaO, and 1.0% Na2O.