Labware
Corning Cell Culture Consumables
Corning Logo
 Liquid Handling | Microplate Selection | All Our Cell Culture Workflow Solutions
We are proud to offer Corning products as a part of our comprehensive offering for all your cell culture needs.
Corning cell culture products offer some of the most advanced surface treatments and culture formats on the market. As a Premier Supply Partner of Corning Life Science products, we offer a portfolio of Corning Cell Culture products including: cell culture flasks, cell culture dishes, pipettes, spinner flaks, centrifuge tubes, microplates, roller bottles, cryogenic vials and culture tubes.
Corning Cell Culture Product Categories:
Corning Low Profile Flask Compared with T175 Flask
Corning Low Profile Flask Compared with T175 Flask
  Corning Cell Culture Flasks | All Plastic Cell Culture Flasks
  • Cell growth areas from: 25 cm² to 1740 cm²
  • Innovative surface treatments: CellBIND, tissue-culture, ultra low attachment, non-treated,
  • Specialty flasks: low profile, HYPERFlask, RoboFlask
Corning Petri Dish with Grid
Corning Petri Dish with Grid
  Corning Cell Culture Dishes | All Cell Culture Dishes
  • Manufactured from optically-clear virgin polystyrene
  • Innovative surface treatments: Ultra-Web, CellBIND, ultra-low attachment, tissue culture treated
Stripette Serological Pipettes
Stripette Serological Pipettes
  Corning Pipettes | All Pipettes
  • Aspiration, Bacteriological, Pasteur, Serological, and Volumetric pipettes
  • Corning Stripette® brand Serological Pipettes
  • Anti-drip tips
  • Individual wrapped, bulk packaged, or new clean room pack style
  • Sterile
Disposable Spinner Flask 500 mL
Disposable Spinner Flask 500 mL
  Spinner Flasks
  • Plastic disposable spinner flasks from 125 mL to 3 L capacity
  • Reusable Pyrex spinner flasks from 125 mL to 36 L capacity
  • Available with baffles, extra-wide mouth and vertical or angled side-arms
CellSTACK Culture Chamber 10-Stack
CellSTACK Culture Chamber 10-Stack
  CellSTACK® Culture Chambers
  • Cell growth areas from 636 cm² (1 stack) to 25440 cm² (40 Stacks)
  • Surface treatments include: CellBIND, tissue culture treated, and ultra-low attachment surface
Centrifuge Tubes
Centrifuge Tubes
  Centrifuge Tubes
  • Centrifuge tubes in Pyrex glass, polyethylene terephthalate (PET) and polypropylene
  • Plug seal, screw cap or Centristar cap styles
Spatulas and Microspatulas
Spatulas and Microspatulas
  Spatulas and Microspatulas
  • Designed for contamination-free samples
  • Sterile, RNase/DNase free, non-pyrogenic and antistatic
1536 Well Microplates
1536 Well Microplates
  Multiple Well Plates
  • 6 to 1536 well microplates
  • Surface treatments: Ultra-Web, non-binding, low-binding, non-treated, tissue culture treated, CellBIND, poly-D-lysine
  • Available plate styles: low volume, UV, Filtex, PCR, Protein Crystallization, and storage
1700cm2 Expanded Surface Roller Bottle
1700 cm² Expanded Surface Roller Bottle
  1700 cm² Expanded Surface Roller Bottle
  • Cell growth areas range from 800 cm² to 1750 cm²
  • Cap Styles: Easy Grip, Easy Grip Vented
  • Surface treatments: CellBIND, tissue culture treated
  • Printed lot numbers,
  • Sterile and non-pyrogenic
1000 mL Erlenmeyer Flask
1000 mL Erlenmeyer Flask
  Erlenmeyer Flasks
  • Ideal for shaker culture applications, liquid handling or storage
  • Bottle capacities: 125 mL to 3000 mL
  • Available with or without baffles
  • 26 mm to 46 mm neck diameter
  • Cap styles: plug seal or vented caps
  • Sterile
Cryogenic Vials
Cryogenic Vials
  Cryogenic Vials
  • Vial capacity: 1.2 mL to 5.0 mL
  • External thread, internal thread, or plug seal caps
  • Packaged in boxes, racks or trays
Culture Tubes
Culture Tubes
  Culture Tubes
  • Cell culture treated tubes supplied racked
  • Sterilized by gamma radiation
  • Threaded plug seal caps prevent leakage
Back to Top