Glassware Catalog
Reducing/Enlarging Adapters
Connecting  •  Reducing/Enlarging  •  Septum-Inlet  •  Specialty  •  Tubing  •  Vacuum
On this page: Reducing/enlarging adaptersBushing-type reducing adapters
Offset reducing/enlarging adaptersFlexible PTFE adaptersAccessories
Reducing/enlarging adapters
Reducing/enlarging apdaters
Top
S.T. Joint
Bottom
S.T. Joint
Catalog Number
Reducing
14/20
24/40
19/22
24/40
24/40
29/32
24/40
34/45
24/40
45/50
24/40
55/50
24/40
65/40 (ball)
24/40
75/50 (ball)
34/45
55/50
34/45
65/40 (ball)
34/45
75/50 (ball)
45/50
55/50
45/50
65/40 (ball)
45/50
75/50 (ball)
55/50
65/40 (ball)
55/50
75/50 (ball)
Enlarging
24/40
14/20
24/40
19/22
29/32
24/40
34/45
24/40
45/50
24/40
55/50
24/40
Back to Top
Bushing-type reducing adapters
Top
S.T. Joint
Bottom
S.T. Joint
Catalog Number
24/40
55/50
24/40
65/40 (ball)
24/40
75/50 (ball)
Back to Top
Offset reducing/enlarging adapters
Offset reducing/enlarging adapters
Top
S.T. Joint
Bottom
S.T. Joint
Catalog Number
Reducing
14/20
24/40
19/22
24/40
24/40
29/32
24/40
34/45
Enlarging
24/40
14/20
24/40
19/22
29/32
24/40
34/45
24/40
Back to Top
Flexible PTFE adapters
The flexible corrugated PTFE tubing is pliable enough to create an offset adapter.
Inner
S.T. Joint
Inner
S.T. Joint
Catalog Number
14/20
14/20
19/22
19/22
24/40
14/20
24/40
19/22
24/40
24/40
24/40
29/32
29/32
29/32
Flexible PTFE adapters
Outer
S.T. Joint
Inner
S.T. Joint
Catalog Number
14/20
14/20
14/20
24/40
19/22
19/22
19/22
24/40
24/40
14/20
24/40
19/22
24/40
24/40
24/40
29/32
29/32
24/40
29/32
29/32
Back to Top
ACCESSORIES
Fritted adapters
Extra-coarse fritted disc prevents solids from being pulled into condenser when removing solvents from solid-solvent mixtures.
Fritted adapters
Outer
S.T. Joint
Inner
S.T. Joint
Catalog Number
24/40
14/20
24/40
24/40
29/32
14/20
29/32
24/40
29/32
29/32
Back to Top
Splash protectors
Unique design has two baffles with horizontally opposing holes that may also be used with other distillation techniques.
Splash protectors
Outer
S.T. Joint
Inner
S.T. Joint
Catalog Number
24/40
14/20
24/40
24/40
29/32
14/20
29/32
24/40
29/32
29/32
Back to Top
See the Evaporator Section for a full listing of Splash-Guards