Glassware Catalog
Condensers
On this page:
Super Condensers | Triple-jacket Coil Condenser | Triple-coil Condensers | Airflux Condenser Apparatus | Coiled Reflux Condensers | Sigma-Aldrich® Jacketed Friedrichs Coil Condensers | Sigma-Aldrich® Modified Friedrichs Condensers | Dry Ice Condensers | Rotary Evaporator Condenser | Cold-finger Condensers | Graham Condensers | Hartzler Condensers | Short Condenser | Liebig Condensers | Modified Davies Condensers
Super Condensers
Excellent for volatile compounds.
Combines all of these condenser features:
Cold-finger • Coiled • Liebig • Vigreux
Overall H
(cm)
Surface area
(cm2)
O.D.
(mm)
S.T. 24/40 Joints
Product No.
S.T. 29/32 Joints
Product No.
32 405 42 Z224979 Z517402
37 510 42 Z224987 Z517410
41 610 42 Z224995 Z517429
51 820 42 Z225002 Z517437
back to top
Triple-jacket Coil Condenser
Triple jacketing provides maximum cooling surface area. Minimizes loss of volatile components.
Excellent for low-boiling solvents.
Does not flood as readily during reflux surges.
Overall H
(cm)
Surface area
(cm2)
O.D.
(mm)
S.T. 24/40 Joints
Product No.
S.T. 29/32 Joints
Product No.
44 1,230 60 Z422274 Z517445
back to top
Triple-coil Condensers
Large surface area provides efficient condensation of refluxing liquids with reduced risk of flooding.
For use with flasks up to 72L.
Efficient, high-volume condensation for large system applications.
Overall H
(cm)
Surface area
(cm2)
O.D.
(mm)
Ball Socket Joints Product No.
Single Stack, 43cm coil
61 3,650 75 65/40 Z421928
61 3,650 75 75/50 Z421936
Double Stack, two 43cm coil sets
107 7,015 75 65/40 Z421944
107 7,015 75 75/50 Z421952
back to top
Airflux Condenser Apparatus
Closed coolant system eliminates wastewater and accidental laboratory flooding. Ideal for solvents boiling between 60 and 150 °C.
Efficient aluminum heat sink disperses heat. Remove plastic cap on heat sink to fill with distilled water. Water passes through to glass condenser. Complete apparatus includes Airflux aluminum heat sink, removable glass condenser, and boiling flask.
  • Use with flasks up to 250 mL size and heating mantles with 60 W max. output
  • Condensers can be mounted in tandem* for flasks sizes up to 1 L and 300 W heating mantles with a controller for steady reflux rate
  • Mounts to 1/2 in. lattice-rods or support stands via built-in clamp on heat sink
Airflux
size
Flaskcap.
(mL)
S.T. 24/40 joints
Product No.
S.T. 24/29 joints
Product No.
S.T. 29/32 joints
Product No.
Small 100 Z410365 Z410381 Z410411
Large 250 Z410373 Z410403 Z410438
*Note: Do not use tandem mounted water-cooled Airflux units to condense solvents boiling over 100 °C. The lower unit works at a temperature close to the boiling point of the solvent at high reflux rates. Alternative coolants such as ethylene glycol may be suitable. Tests should be carried out to determine satisfactory performance.
back to top
Coiled Reflux Condensers
The coiled reflux condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by
allowing vapors to condense on the inner wall of the jacket.
For use with dry ice/acetone slush or liquid nitrogen in cold-finger.
Overall H
(cm)
S.T. Joints Standard
Product No.
Jacketed
Product No.
10* 14/20 Z531073 Z531227
20 14/20 Z531081 Z531235
20 24/40 Z531103 Z531243
20 29/32 Z531111 Z531251
30 14/20 Z531138 Z531278
30 24/40 Z531146 Z531286
30 29/32 Z531154 Z531294
40 24/40 Z531162 Z531308
40 29/32 Z531170 Z531316
50 24/40 Z531189 Z531324
50 29/32 Z531197 Z531332
60 24/40 Z531200 Z531340
60 29/32 Z531219 Z531359
* Jacket length
back to top
Sigma-Aldrich® Jacketed Friedrichs Coil Condensers
The molded spiral condensing tube around the inner coil creates additional surface area for improved
efficiency. 54mm O.D.
Overall H
(cm)
S.T. 24/40 Joints
Product No.
S.T. 29/32 Joints
Product No.
26 Z517232 Z517240
37 Z517259 Z517267
back to top
Sigma-Aldrich® Modified Friedrichs Condensers
Molded spiral condensing coolant tube fits closely within the jacket to force vapors along the spiral path.
Features:
  • Modified feed tube to reduce chance of breakage
  • Additional sizes to meet specific applications
Jacket O.D.
(mm)
Overall H
(mm)
S.T. Joint Product No.
25 230 14/20 Z517143
37 160 14/20 Z517151
37 270 24/40 Z517178
37 270 29/32 Z517186
37 370 24/40 Z517194
37 370 29/32 Z517208
54 300 24/40 Z517216
54 300 29/32 Z517224
back to top
Dry Ice Condensers
For Büchi rotary evaporators R110, R114, R124 and R151.
For use with short-stem duct. 35/25 ball joint.
Description Product No.
Trap with hose barb Z117242
Trap with high vacuum, hollow glass stopcock Z422282
35/20 ball joint. Modified feed tube allows recharging during stripping.
Z152447
With S.T. 24/40 ground-glass joints.
Z103098
For use with dry ice/acetone slush or liquid nitrogen in cold-finger.
back to top
Rotary Evaporator Diagonal Condenser
35/25 side ball joint at 105° angle, and No. 50 flange. 37 cm overall length.
Z224774
back to top
Cold-finger Condensers
31 cm overall length. 25 L x 6 cm diam. cold-finger.
For reactions using low-boiling solvents.
S.T. Joint Product No.
14/20 Z164046
24/40 Z164038
29/32 Z517453
back to top
Graham Condensers
The Graham condenser has a large cooling surface area by allowing coolant to circulate around the inner coil.
For use with dry ice/acetone slush or liquid nitrogen in cold finger.
Overall H
(cm)
S.T. Joints Standard
Product No.
Jacketed
Product No.
10* 14/20 Z531073 Z531227
20 14/20 Z531081 Z531235
20 24/40 Z531103 Z531243
20 29/32 Z531111 Z531251
30 14/20 Z531138 Z531278
30 24/40 Z531146 Z531286
30 29/32 Z531154 Z531294
40 24/40 Z531162 Z531308
40 29/32 Z531170 Z531316
50 24/40 Z531189 Z531324
50 29/32 Z531197 Z531332
60 24/40 Z531200 Z531340
60 29/32 Z531219 Z531359
* Jacket length
back to top
Hartzler Condensers
34 cm column length. Lower primary condenser of bulbous portions connected with each other and with an upper curvilinear
column. Designed by Harold E. Hartzler, Goshen, IN.
S.T. Joint Product No.
19/38 Z175889
24/40 Z175897
29/42 Z175900
back to top
Short Condenser
With S.T. 24/40 ground-glass joints. Useful in distilling solids. May be used for conventional distillations as a condenser or a
jacketed column. 17.5 cm jacket length.
Z115754
back to top
Liebig Condensers
Condensers have a standard taper outer joint and a lower inner drip-tip joint.
Hood space constraints? Choose a condenser by its overall height.
Overall H
(cm)
S.T. Joints Standard
Product No.
6* 14/20 Z530948
10* 14/20 Z530956
20 14/20 Z122416
20 24/40 Z530964
20 29/32 Z530972
30 14/20 Z530980
30 24/40 Z530999
30 29/32 Z531006
40 24/40 Z531014
40 29/32 Z531022
50 24/40 Z531030
50 29/32 Z531049
60 24/40 Z531056
60 29/32 Z531065
* Jacket length
back to top
Modified Davies Condensers
Additional internal cooling surface increases efficiency with negligible hold-up when used at a standard 75° angle.
Jacket L
(cm)
S.T. 24/40 Joint
Product No.
S.T. 29/32 Joint
Product No.
25 Z214485 Z517461
40 Z214825 Z517488
back to top