Glassware Catalog
Microscale Glassware Kits
Metals/Metal Hydrides Test Kit  •  All-In-One  •  Corning Organic Chemistry  •  Microscale Kits
On this page:
ACE microscale glassware kitsKONTES Williamson microscale glassware kits
ACE microscale glassware kits
Designed to give students experience with a wide range of organic reactions with increased safety. Kits contain microscale glassware needed to perform experiments described in the textbook Microscale Organic Laboratory. Greaseless, threaded S.T. 14/10 connections can be interfaced with traditional S.T. 14/10 joints. Conical reaction vials include open-top caps, septa, and O-rings. Glassware in the Deluxe kit is supplied in a foam-lined plastic carrying case. The BASIC kit is supplied in a clear plastic, compartmentalized box with tilt lid.
Ace kits are available in two configurations to meet specific requirements:
  • MICRO/MINI-Lab kit Deluxe – covers all 52 experiments in the textbook
  • MICRO/MINI-Lab BASIC KIT II – covers 95% of the experiments in the textbook as well as the majority of experiments in Organic Laboratory Techniques
Deluxe kit – Z174831
Basic kit – Z245240
Components Deluxe
Qty in kit
Basic
Qty in kit
Replacement parts
A. Air condenser
1
-
B. Claisen adapter
1
1
C. Conical reaction vial, 0.1mL
2
-
-
D. Conical reaction vial, 3.0mL
2
1
E. Conical reaction vial, 5.0mL
1
1
F. Craig recystallization tube, 1mL
1
-
G. Craig recystallization tube, 2mL
1
1
H. Drying tube
1
-
I.  Gas-delivery tube
1
-
J. GC collection tube, 5/5M
2
-
K. Hickman column
-
1
-
L. Hickman still head
1
-
M. Jacketed condenser
1
1
N. Multi-purpose adapter
-
1
O. Round-bottom flask, 10mL
-
1
P. Sample syringe
1
-
-
Q. Spin vane
-
1
Special pricing is available for all orders of 20 or more microscale kits.
Please contact your local sales representative for a quotation.
back to top
KONTES Williamson microscale glassware kits
Kits contain glassware needed to perform experiments described in the textbook Macroscale and Microscale Organic Experiments. These economical microscale glassware kits have a unique, positive connection system without the use of threads or S.T. joints. Glassware is supplied in a foam-lined Ekono-Case.
Standard kit A – Z256617
Standard kit B – Z256625
Basic kit – Z256633
Components Standard kit A
Qty in kit
Standard kit B
Qty in kit
Basic Kit
Qty in kit
A. Centrifuge/sublimation tube, 15mL
1
1
1
B. Chromatography column, 150mm
1
1
1
C. Connecting adapter
1
1
1
D. Connector only
1
-
-
E. Connector only, Viton
1
1
1
F. Connector w/support rod
1
1
1
G. Distillation column
1
1
1
H. Distillation head, 60mm
1
1
1
I. Filter adapter, Pluro stopper
1
1
1
J. Flask, Erlenmeyer, 10mL
3
4
-
K. Flask, filtering, 25mL
1
1
1
L. Flask, long-neck, 5mL
1
1
-
M. Flask, short-neck, 5mL
1
1
1
N. Hirsch funnel, w/20 µm PE disc
1
1
1
O. Pipette, 1mL
1
-
-
P. Reaction tube, 10 x 100mm
5
4
2
Q. Sleeve stopper, 8mm
2
2
-
R. Spatula, SS
1
1
-
S. Stirring bar, 4 x 12mm
2
-
-
T. Stopcock, one-way, PE/nylon
1
1
1
U. Syringe, PE
1
1
1
V.Thermometer adapter
1
1
1
W. Tubing, PTFE, 2ft (fits 20 gauge needle)
1
1
1
back to top