Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glassware Catalog
Heating Mantle Controllers - Sigma-Aldrich Glassware Catalog
Glas-Col Series O  •  Glas-Col Series TM  •  Glas-Col Controllers
On this page:
Cordtrol IIPowrTrol heat controllersDigiTrol II heat controllersRampTrol controllers
Cordtrol II
Detachable heating mantle control. Solid-state. Features non-arcing circuitry for safety. Has high-ratio vernier dial for precise setting, exacting repeatability, and expanded range. Compact (3 3/4 L x 2 1/2 W x 2 3/4 in. H) and encased in anodized aluminum. Has no-mar rubber feet and SS bracket (for 1/2in. diameter rods) for mounting on a lab bench, distillation rack, or mantle. Can be used with metal-housed heating mantles of up to 900W. 120V, 5A.
Z10,769-7
Back to top
PowrTrol heat controllers
A solid-state, proportional-voltage power control that provides precise manual control of mantles and tapes. The output is adjustable from 3 to 100% of rated voltage. Can be mounted to a lattice or used on bench top. CE compliant.
Volts
Amps
Watts
Catalog Number
120
10
1,200
240
10
2,400
Back to top
DigiTrol II heat controllers
  • Microprocessor based temperature control
  • Accuracy: 0.1% of span
  • Range: 32 to 999°F, 0 to 750°C
  • CE compliant
A microprocessor controller to be used with mantles, tapes, small ovens, and cords. The control features a digital display for set point and actual temperature. Easy set-up with selectable °F/°C and up and down temperature set point keys. Includes a 6in., stainless steel immersion thermocouple.
Volts
Amps
Watts
Catalog Number
120
15
1,800
240
10
2,400
Back to top
RampTrol controllers
  • Single output
  • Optional retransmit of set point or process variable
  • Accuracy: 0.1% of span, temperature ±0.2%
  • 8 W x 8 D x 4 3/4 in. H
  • Not CE compliant
A microprocessor controller provides ramping operation in four files with six steps, or non-ramping set-point operation. Multiple programming options, with serial port for PC communication.
Thermocouple Type
Range (°C)
120V
Catalog Number
240V
Catalog Number
J
0 - 816
K
-200 - 1,371
T
-200 - 399
Back to top