Glassware Catalog
Vacuum Valves - Sigma-Aldrich Glassware Catalog
Manometers/Gauges  •  Valves
On this page:
Adjustable rapid-purge valveAldrich SAFE-PURGE valvesFirestone valve
Adjustable rapid-purge valve
Compact design allows for easy mounting anywhere in a vacuum system. For use when purging an evacuated apparatus with inert gas. Two-piece PTFE plug with Viton O-ring and SS spring is adjustable for pressures greater than 5 psi.
Back to top
Aldrich SAFE-PURGE valves
The Aldrich SAFE-PURGE valves provide for safe, high-performance operation. Both the manual and automatic spring-loaded styles feature a durable PTFE metering valve.
Manually adjustable PTFE valve
Used with purge gas pressure under 5 psi.
Automatic spring-loaded PTFE valve
Used with purge gas pressure greater than 5 psi. Should any oil pass the check valve, the spring-loaded plug will prevent any oil or air from entering the system
Please refer to Aldrich Technical Information Bulletin AL-198 for procedural details.
Back to top
Firestone valve
Connect reaction vessel, house-vacuum, and purge gas to the Firestone valve via 10mm o.d. connections. A rapid and efficient purge valve for 100% replacement of air in reaction vessels. Automatically controls gas flow and pressure. Conserve expensive gases by decreasing flow to almost zero after purging.
Back to top