Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Hettich Selection Guide
Mikro-220R (115 Volts)
Hettich Product No. 2205-01
Sigma-Aldrich Product No. Z724076
Select an available tube type below to view volume, tube quantity and centrifuge accessory information
Available Tube types:
Blood / Urine Conical Conical / Skirted Culture Plate Microcentrifuge Microtiter Plate PCR Strips Round Bottom
Blood / Urine
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
9.0-10.0 6 1016 Z601381 1635 Z601152 N/A N/A
10.0 6 1016 Z601381 1635 Z601152 N/A N/A
1.1-1.4 12 1015 Z722758 1054-A Z724408 N/A N/A
2.6-2.9 12 1015 Z722758 1054-A Z724408 N/A N/A
2.7-3.0 12 1015 Z722758 1054-A Z724408 N/A N/A
4.9 12 1015 Z722758 N/A N/A N/A N/A
4.5-5 12 1015 Z722758 N/A N/A N/A N/A
4.0-7.0 12 1015 Z722758 N/A N/A N/A N/A
7.5-8.2 12 1015 Z722758 N/A N/A N/A N/A
9.0-10.0 12 1015 Z722758 N/A N/A N/A N/A
10.0 12 1015 Z722758 N/A N/A N/A N/A
Conical
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
15.0 6 1016 Z601381 1631 Z601160 N/A N/A
50.0 6 1016 Z601381 1641 Z601179 N/A N/A
15.0 12 1015 Z722758 N/A N/A N/A N/A
Microcentrifuge
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
0.2 30 1189-A Z623482 2024 Z601209 N/A N/A
0.4 30 1189-A Z623482 2024 Z601209 N/A N/A
0.5 30 1189-A Z623482 2023 Z601195 N/A N/A
0.8 30 1189-A Z623482 2023 Z601195 N/A N/A
1.5 30 1189-A Z623482 2031 Z652229 N/A N/A
2.0 30 1189-A Z623482 N/A N/A N/A N/A
0.2 24 1195-A Z623490 2024 Z601209 N/A N/A
0.4 24 1195-A Z623490 2024 Z601209 N/A N/A
0.5 24 1195-A Z623490 2023 Z601195 N/A N/A
0.8 24 1195-A Z623490 2023 Z601195 N/A N/A
1.5 24 1195-A Z623490 2031 Z652229 N/A N/A
2.0 24 1195-A Z623490 N/A N/A N/A N/A
0.2 6 1161 Z601349 1378 Z601357 N/A N/A
0.4 6 1161 Z601349 1378 Z601357 N/A N/A
0.5 6 1161 Z601349 1379 Z601365 N/A N/A
0.8 6 1161 Z601349 1379 Z601365 N/A N/A
1.5 6 1161 Z601349 1377 Z601373 N/A N/A
2.0 6 1161 Z601349 1377 Z601373 N/A N/A
0.2 48 1158-L Z651664 2024 Z601209 N/A N/A
0.4 48 1158-L Z651664 2024 Z601209 N/A N/A
0.5 48 1158-L Z651664 2023 Z601195 N/A N/A
0.8 48 1158-L Z651664 2023 Z601195 N/A N/A
1.5 48 1158-L Z651664 2031 Z652229 N/A N/A
2.0 48 1158-L Z651664 N/A N/A N/A N/A
0.2 24 1154 Z722731 2024 Z601209 N/A N/A
0.4 24 1154 Z722731 2024 Z601209 N/A N/A
0.5 24 1154 Z722731 2023 Z601195 N/A N/A
0.8 24 1154 Z722731 2023 Z601195 N/A N/A
1.5 24 1154 Z722731 2031 Z652229 N/A N/A
2.0 24 1154 Z722731 N/A N/A N/A N/A
0.2 12 2218-A Z732001 2024 Z601209 N/A N/A
0.4 12 2218-A Z732001 2024 Z601209 N/A N/A
0.5 12 2218-A Z732001 2023 Z601195 N/A N/A
0.8 12 2218-A Z732001 2023 Z601195 N/A N/A
1.5 12 2218-A Z732001 2031 Z652229 N/A N/A
2.0 12 2218-A Z732001 N/A N/A N/A N/A
Round Bottom
Volume (mL) Tube Quantity Hettich Rotor
Product No.
Sigma-Aldrich Rotor
Product No.
Hettich Adaptor
Product No.
Sigma-Aldrich Adaptor
Product No.
Hettich Bucket
Product No.
Sigma-Aldrich Bucket
Product No.
15.0 6 1016 Z601381 1635 Z601152 N/A N/A
50.0 6 1016 Z601381 N/A N/A N/A N/A
5.0 12 1015 Z722758 1054-A Z724408 N/A N/A
15.0 12 1015 Z722758 N/A N/A N/A N/A