Spectral Viewer
FT-IR Library: an essential reference for research
The FT-IR Library is an essential reference for:
  • Researchers and technicians in chemistry and biochemistry
  • Undergraduate and graduate students
  • Teachers and instructors
  • Spectroscopists
  • Scientific libraries
Features:
  • Displays FT-IR spectra in % Transmittance or Absorbance along with chemical structure and technical data
  • Text Searching by Chemical Name, Catalog Number, CAS Number, and Molecular Formula
  • Advanced data field searching on Chemical Category and physical properties such as Boiling Point, Melting Point, Density, Flash Point, Molecular Weight, and Refractive Index
  • Spectra processing using Zoom and Peak Labeling
  • Print spectra, structure, technical data, and peak listings
  • Export spectra to a file (Bitmap, Metafile, or JPEG formats) or to the clipboard (Bitmap or Metafile formats)
  • Web link button provides additional technical data from our website
  • Fully compatible with Spectral Viewer FT-NMR libraries - further enhance the utility of this powerful electronic reference with the addition of our FT-NMR libraries
  • Expandable - As new spectral libraries become available they can be added at a minimal fee with no subscription or maintenance fees required
Minimum system requirements:
Pentium 200MHz, Windows 95 / 98 / NT4-SP5 / 2000 / ME / XP, CD-ROM drive, 300MB hard disk space, 64MB RAM
FT-IR 2002 library (11,000 compounds)
Version Cat. No.
Single user, commercial Z54,016-1
Single user, academic Z54,028-5
Network version Z54,039-0
Demo program Z54,159-1
For more information email labequipment@sial.com.
Spectral Viewer is a trademark of Sigma-Aldrich Co. LLC
Chemical Abstracts is a registered trademark of the American Chemical Society
Pentium is a registered trademark of Intel Corp.
Windows is a registered trademark of Microsoft Corp.