Register your Scepter™ 3.0 Cell Counter
Scepter™ 3.0 cell counterWhen you register your Scepter™ 3.0 instrument, you will receive regular updates on your instrument's software and firmware.
While other automated counters consume bench space and rely on object recognition software, manual focusing, and manual sample transfer and loading, the unique Scepter™ 3.0 cell counter provides automated, impedance-based counting without the error of vision-based systems. With its precision-engineered sensor, the Scepter™ cell counter delivers accurate, consistent cell counts in 15-25 seconds, depending on the sensor and the sample.
To ensure that you stay informed regarding software releases and upgrades and application notes, simply register your automated cell counter.
To register your Scepter™ 3.0 instrument, complete and submit the below form.
Fields indicated by an * are required.
Found on back of the instrument
You can unsubscribe at any time