Validated Cell Types for the Scepter™ 3.0 Cell Counter
The Scepter™ counter has been shown to be accurate over a range of cell types and sizes. See below for a list of which mammalian and other key phenotypes have been validated for Scepter™ counting precision.
  Measured    
Cell Type size (μm) 40 μm sensor 60 μm sensor
2102 Ep 15-19   *
454 beads     *
A172 15   *
A253 14-18   *
A375 16   *
A431 15-17   *
A549     *
Algae (various) 7-9 * *
B35 13-16 *  
B Cells 6-11 *  
C2C12 12 * *
C305 12-14 * *
C6 12-13   *
CA46 10-12 * *
Caco-2 17   *
CHO 14-17   *
COS-1 12 *  
Cos-7 15   *
D283 12 *  
Daudi 10-12 * *
DU-145 15-17   *
Epithelia 14-15   *
HCT-116 10 * *
HEK293 11-15   *
HeLa 12-14   *
HepG2 12   *
HFF 18-20   *
Hs27 14 * *
HT-1080 14-16   *
HT-29 11   *
HUH7- Hepatoma line     *
Human ES Cells 9-12 *  
HUVEC 14-15   *
IMR-32 12-14 * *
IMR-90 15 * *
Jurkat 13   *
K562 22   *
KB 14   *
KG-1 10-13 * *
L6 14-16   *
LNCaP 15-16   *
Luminex® beads 5-6 *  
MCF7 15-17   *
MDCK 13-15   *
  Measured    
Cell Type size (μm) 40 μm sensor 60 μm sensor
Meg-01 16-17   *
MG-63 15-17   *
Mouse ES Cell 5-13 * *
Mesenchymal Stem Cell 15-16 *  
MRC-5     *
NCI-H146 10-13 *  
NIH 3T3 15   *
NTERA2, clone D1 13 * *
OK 17-18   *
PBMCs 7-12 *  
PC12 9-13   *
Primary Astrocytes 7 *  
Primary Neuronal Cell   *  
Raji 12-15 * *
Ramos 11-12 * *
Rat Dorsal Root
Ganglion Cells
7 *  
Rat Whole Blood 4.6 *  
Red Blood Cells 5-7 *  
Rat Neural Stem Cell 11-13   *
RAW 264.7 12-15   *
RBL 11-13 * *
RIN-mF5 13-14   *
SF9 13   *
SH-SY5Y 12   *
Sk-Br-3 15-20   *
SK-MEL-28 17-19   *
SK-N-MC 14-15   *
SK-N-SH 14-15   *
Splenocytes 7-9 *  
SW-480 15   *
SW-620 13-14 * *
T84 14-18   *
T98G 17   *
TF-1 13-14 * *
U251 16-20   *
U2OS 16-19   *
U266 12   *
U87-Human
Glioblastoma cell line
12-14   *
U937 11-13 * *
WI-38 12-15   *
Y79 13-14   *
Yeast- Pichia Pastoris 5 *  
Yeast - S. cerevisiae 6 *  
* Recommended based on size         * Customer Validated         * { MilliporeSigma } Validated
Scepter™ 3.0 Handheld Automated Cell Counter Ordering Table
Product Description Qty Catalogue No.
Scepter™ 3.0 Handheld Automated Cell Counter Kit
   Includes Scepter™ 3.0 Handheld Automated Cell Counter,
   50/pk of Scepter™ 3.0 Sensors (40 μm or 60 μm),
   Scepter™ Test Beads, Charging station and mounting kit,
   Quick Start Guide, and Safety information.
         with 40 μm sensors 1 PHCC340KIT
         with 60 μm sensors 1 PHCC360KIT
Scepter™ 3.0 Sensors only, 40 μm 50 PHCC340050
Scepter™ 3.0 Sensors only, 40 μm 250 PHCC340250
Scepter™ 3.0 Sensors only, 40 μm 500 PHCC340500
Scepter™ 3.0 Sensors only, 60 μm 50 PHCC360050
Scepter™ 3.0 Sensors only, 60 μm 250 PHCC360250
Scepter™ 3.0 Sensors only, 60 μm 500 PHCC360500
Scepter™ 3.0 Test Beads only, 5 mL 1 PHCC3BEADS
Scepter™ 3.0 Charging Station only 1 PHCC3CHARG
Scepter™ 3.0 Charging Station Mounting Kit only 1 PHCC3WKIT