Stericup® E and Steritop® E Filtration Systems
Try Stericup® E filtration, and help science become more sustainable
Scientific progress can be resource-intensive—but it doesn’t always have to be, and we’d like to help change that. With Stericup® E and Steritop® E devices, it’s effortless for your lab to reduce plastic use and biohazardous waste while maintaining confidence in your sterile filtration process.  And with every purchase, you automatically pay the green forward, because we’ll contribute to supporting sustainable science on your behalf.
Promotion Details:
For every case (12 units) of Stericup® E or Steritop® E filters purchased, we will donate $10.00 USD towards green lab education, innovation and advocacy.
Maximum donation for this promotion will be $25,000 USD. Donation program active through 12/31/2021.
Stericup Donation Status
Choose Stericup® E and Steritop® E filters based on your volume and thread size requirements:
Product No. Name Pore Size (μm) Thread Size Max Process Volume Membrane/Application Qty/Pk
SEGPU0538 Stericup® E-GP Funnel-less Sterile Vacuum Filtration System 0.22 38 mm 500 mL Millipore Express® PLUS (PES) / fast filtration of tissue culture media and buffers 12
SEGPU0545 Stericup® E-GP Funnel-less Sterile Vacuum Filtration System 0.22 45 mm 500 mL Millipore Express® PLUS (PES) / fast filtration of tissue culture media and buffers 12
SEGPU1138 Stericup® E-GP Funnel-less Sterile Vacuum Filtration System 0.22 38 mm 1000 mL Millipore Express® PLUS (PES) / fast filtration of tissue culture media and buffers 12
SEGPU1145 Stericup® E-GP Funnel-less Sterile Vacuum Filtration System 0.22 45 mm 1000 mL Millipore Express® PLUS (PES) / fast filtration of tissue culture media and buffers 12
SEGPT0038 Steritop® E-GP Funnel-less Sterile Vacuum Filtration System 0.22 38 mm 500 - 1000 mL Millipore Express® PLUS (PES) / fast filtration of tissue culture media and buffers 12
SEGPT0045 Steritop® E-GP Funnel-less Sterile Vacuum Filtration System 0.22 45 mm 500 - 1000 mL Millipore Express® PLUS (PES) / fast filtration of tissue culture media and buffers 12