ELISA Kits
 Enzyme linked immunosorbent assay (ELISA) kits
Detect protein analytes with confidence using our ELISA kits that deliver reliable and reproducible results. Our pre-validated assays allow you to quantify biomarkers at the pg/mL level. Leveraging years of continuous innovation, our offerings consist of a portfolio of >1,800 assays to address your specific needs. From complex sample types and high-throughput screening, we invite you to explore the diverse range of species and biomarker classes available within our portfolio.
  • Over 1,800 ELISAs now available
  • Detection of cytokine markers for all species within our portfolio
  • Validation criteria including: inter- and intra-assay CV, sensitivity, recovery linearity, biological range, cross-reactivity, species-specificity, sample and shipping stability, and method comparison
  • Variability avoidance with accurate reagent dispensing and thorough plate washing
  • All kits backed by our technical resources
Bioguarantee Antibodies
Sigma-Aldrich’s rigorously tested ELISA kits are all backed by our Bioguarantee Program. We invite you to evaluate our antibody products with complete peace of mind.