Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Fluorescent Labeled Nucleotides
Cat. No. Description Recommended Application Pack size
19680 Aminoallyl-dUTP-Atto 425 triethylammonium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
69303 Aminoallyl-XX-dUTP-Atto 488 triethylammonium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
69338 Aminoallyl-XX-dUTP-Atto 532 triethylammonium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
30387 Aminoallyl-dUTP-Atto 550 triethylammonium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
30098 Aminoallyl-dUTP-Atto 647N triethylammonium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
19685 Rhodamine-12-dUTP triethylammonmium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
00294 Fluorescein-12-dUTP triethylammonium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
69383 Aminoallyl-dUTP-Texas Red® triethylammonium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
19518 Aminoallyl-dUTP-Cy®3 triethylammonium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
19475 Aminoallyl-dUTP-Cy®5 triethylammonium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
69302 Propargylamino-dCTP-Cy®3 triethylammonium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
69104 Propargylamino-dCTP-Cy®5 triethylammonium salt solution Incorporation into DNA/cDNA by PCR with Taq polymerase in-house data and Nick Translation with DNAse I/ DNA Polymerase I 10 nmol
19511 Mant-ADP triethylammonium salt solution  studies of: MYOSIN-ATPase cycle; Dissociation kinetic proteinkinase A;  FRET: kinesin; Conformational dynamic: DnaC-protein 1.5micromol
18723 Mant-ATP triethylammonium salt solution Conformational dynamic of DnaB/C-protein and Csk; Fluorescence stop-flow kinetics: PKA; FRET; AC, myosin 1.5micromol
69258 Mant-dATP triethylammonium salt solution Conformational dynamic  studies; Enzymatic kinetic studies 0.5micromol
69244 Mant-GDP triethylammonium salt solution Conformational dynamic of DnaB/C-protein and Csk; Fluorescence stop-flow kinetics: release factor, elF5B, EF-
1B, EF-Tu
1.5micromol
69234 Mant-GTP triethylammonium salt solution Inhibition of AC-isoform and GTPs; Activity measurement: GC; Specifity measurements with isoforms of Acs; FRET: AC, edema factor 1.5micromol
69233 Mant-dGTP triethylammonium salt solution Resonance energy transfer studies (FRET); Enzyme kinetic studies 0.5 micromol
69299 TNP-GTP triethylammonium salt Conformational dynamic  studies; Enzymatic kinetic studies 1microm