Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Hematology & Histology
Hematoxylin
Hematoxylin is the most widely used natural dye and is extracted from the heartwood of the logwood tree. The tree is native to Central America and extracts were used for dying purposes over 500 years ago. Various formulations of the dye, such as Mayer's, Gill's and Harris' have been used to stain nulcei, mitochondria, mucin, elastic fibers, muscle, collagen, myelin sheaths and phospholipids. Proper staining hematoxylin solutions require oxidation of the dye followed by linking to a metal mordant, typically aluminum.
Products
Certified Hematoxylin