Research Pathology Products
Tissue processing for research pathology requires high quality reagents optimized for histological staining and immunohistochemistry (IHC). For outstanding contrast, color, clarity, and antibody specificity, choose from our broad selection of quality consumables, processing reagents, stains, and research-grade antibodies tested in IHC.
In addition, we offer the SNAP i.d.® 2.0 System for IHC and Western blotting. This system streamlines IHC by enabling simultaneous processing of up to 24 slides, using a controlled vacuum force that removes solutions evenly from all slides at once – in seconds.
 Products for Reliable Tissue Staining & IHC
Tissue Staining Workflow
Featured Solutions
Research-grade Pathology Antibodies
Our portfolio of 18,000 Prestige® Antibodies includes hundreds of routinely used research pathology antibodies. For details on antibody host, specificity, clone (if applicable), visit each product detail page.
SNAP i.d.® System for IHC
Driven by vacuum, this system streamlines both IHC and Western blotting.
 Video Demonstration: SNAP i.d.® System for Immunohistochemistry (IHC)
Watch this video to learn how the SNAP i.d.® system enables processing of up to 24 slides at a time, reducing slide handling and potential variability. The system works with both formalin-fixed, paraffin embedded (FFPE) and fresh, frozen tissue samples.
 Product Information
Description Product No. Quantity per Pack
Base System SNAP i.d.® 2.0 Base SNAP2IHC Not yet available on click to request information
SNAP i.d.® 2.0 Immunohistochemistry Frame SNAP2FRIHC 1
SNAP i.d.® 2.0 IHC Slide Holder SNAP2SH 24