β Amyloid ELISAs
Product Name |
Product Code
|
β-Amyloid 1-40 ELISA, human |
|
β-Amyloid 1-42 ELISA, human |
|
β-Amyloid peptide (Aβ), a 40 to 43 amino acid peptide, is an early and critical feature of Alzheimer’s disease (AD). It is the major protein component of the protein deposits in neurofibrillary tangles, neuritic plaques, and neurophil threads in the cerebral cortex of AD and Down syndrome patients. The increased release of Aβ42/Aβ43 leads to the Aβnormal deposition of human Aβ and the associated neurotoxicity in the brains of affected individuals.1, 2
Solid phase human Aβ40 and human Aβ42 ELISAs are designed for in vitro quantitative detection of human Aβ peptides in sera, cell culture supernatants and EDTA plasma and CSF. The assays recognize both natural and recombinant forms of human Aβ. The detection antibodies used in these kits are capAβle of selective differentiation between human Aβ40 and Aβ42/Aβ43. In both assays, an antibody specific for the NH2-terminus of human Aβ is coated onto the wells of a multiwell plate and binds human Aβ 1-40 or 1-42 present in samples or standards. A second antibody specific for the 1-40 or 1-42 sequence of human Aβ binds to the immobilized protein. After substrate addition, the reaction is visualized at 450 nm in the multiwell plate reader. The intensity of the color is directly proportional to the concentration of the human Aβ present in sample or standard. The assays are highly sensitive (detect human Aβ below 10 pg/mL), specific, fast (6.5 hours total time) and simple (offer precoated plates, all incubations proceed at room temperature and the reagents are supplied ready to use).
Features and Benefits Size
Sensitive
- Highly sensitive (below 10 pg/ml)
Specific
- Specifically measures Aβ40 or Aβ 42/43
- hAβ40 ELISA no cross-reactivity with hAβ42 or hAβ43 up to 10,000 pg/ml
- hAβ42 ELISA no cross-reactivity with hAβ40 or hAβ43 up to 10,000 pg/ml
Quantitative
- All results are quantitative
- Standard concentration curve is run in each assay
Simple
- Precoated plates eliminate the coating step
- All incubations proceed at room temperature
- Any commercially available microplate reader may be used
Fast
Economical
- Reagents may be used for multiple kit runs
- Cost per test is low
Performance Characteristics
- Precision
- Very low intra-assay variations from 1.1% to 3.6%
- Very low inter-assay variations from 1.7% to 3.7%
- Linearity of dilution – correlation coefficient for various dilutions was 0.99 for both assays
- Recovery – average recovery: sera 100%, EDTA plasma 92.8%, cerebrospinal fluid (CSF) 98%, cell culture supernatants 101-109%
Storage
References
- Sambamurti, K., et al., Neuromolecular Med., 1, 1-31 (2002).
- Lorenzo, A., et al., Nat. Neurosci., 3, 460-464 (2000).
- Vassar, R., et al., Science, 286, 735-741 (1999).
back to top
Apoptosis Made Easy Kit
Product Name |
Product Code
|
Apoptosis Made Easy Kit |
|
Caspase 3 Fluorometric Assay Kit |
|
Features and Benefits
- High purity reagents
- CHX and ActD can also be used as tools to investigate the apoptosis inhibitors in these cells
back to top
Flow Cytometry Kit for Apoptosis
Product Name |
Product Code
|
Flow Cytometry Kit for Apoptosis |
|
Annexin V-FITC Apoptosis Detection Kit |
|
Features and Benefits
- Stronger signal and improved detection as compared with FITC, biotin or digoxigenin-labeled Br-dUTP
- Better detection than fluorochrome (fluorescein or BODIPY)-conjugated deoxynucleotides
- Differentiation of non-fragmented DNA with propidium iodide/RNase A solution included in the kit
- Improved identification of non-apoptotic cells with fixed negative control cells included in the kit
- Sufficient for 60 cell suspensions
back to top
Cathepsin Kits
Product Name |
Product Code
|
Cathepsin Detection Assay (Cathepsin L Detection Assay) |
|
Cathepsin Detection Assay (Cathepsin K Detection Assay) |
|
Cathepsin Detection Assay (Cathepsin B Detection Assay) |
|
Features and Benefits
- Fluorogenic substrates easily penetrate the cell membrane and the membranes of the internal cellular organelles
- athepsin intracellular localization identified and visualized by varying the duration and concentration of exposure to the fluorophore MR-cathepsin substrate
- Potent, cell-permeable and non-toxic fluorophore MagicRed™ reagent
- No need for cell lysis nor membrane permeabilization
- Kit could be run on fluorescence microscopes, fluorescence plate readers and flow cytometers
- Works on frozen cells
- Lysosomes and other intracellular organelles identified with acridine orange (AO) included in the kit
- Cell nuclei labeled with Hoechst stain included in the kit
back to top
Cytochrome c Oxidase Assay Kit
Product Name |
Product Code
|
Cytochrome c Oxidase Assay Kit |
|
Mitochondria Isolation Kit |
|
Features and Benefits
- Detects mitochondrial outer membrane integrity
- Detects mitochondrial subcellular fractions
- Use with Sigma Mitochondrial Isolation Kit
- Suitable for 100 tests
back to top
Cytochrome P450 Fluorescent Detection Kits
Product Name |
Product Code
|
Cytochrome P450 Fluorescent Detection Kit (Detects CYP1A1 and CYP1A2) |
|
Cytochrome P450 Fluorescent Detection Kit (Detects CYP2B1/2/4) |
|
Features and Benefits
- Standard and substrates in solution ready to use
- Positive control provided
- Reagents for cell extraction provided
- Suitable for 50 assays
back to top
Leukotriene and Prostanoid Enzyme Immunoassay (EIAs) Assays
Product Name |
Product Code
|
Cysteinyl Leukotriene EIA |
|
Leukotriene B4 EIA |
|
Prostaglandin E1 EIA |
|
Prostaglandin E2 EIA |
|
Substance P EIA |
|
Thromboxane B2 EIA |
|
Features and Benefits Specific
- Specifically measures for each target lipid
- Negligible cross-reactivity with 20-25 related eicosanoid compounds tested (from <0.1 to 2%)
Quantitative
- Standard curve is run in each assay
Simple
- Precoated plates eliminate the coating step
- All incubations proceed at room temperature
- Any commercially available multiwell plate reader may be used
- Reagents are color coded
Fast
Economical
- Reagents may be used for multiple kit runs
Performance Characteristics
- Low intra- and inter-assay variability
- Linear standard curve
- Recovery – ranges from 99-111%
back to top
ELISA Kits
Product Name |
Product Code
|
Akt/PKB ELISA |
|
Amyloid Precursor Protein ELISA Human |
|
Human β-Amyloid ELISA Kit (Human β Amyloid 1-40) |
|
Human β-Amyloid ELISA Kit (Human β Amyloid 1-42) |
|
ATF2 ELISA |
|
Bcl-2 ELISA Human |
|
CREB ELISA |
|
ERK 1&2 ELISA |
|
Epidermal Growth Factor Receptor ELISA, Human |
|
Heat Shock Protein 27 ELISA, Human |
|
IκBαELISA Human |
|
Insulin Receptor β Subunit ELISA |
|
JNK 1&2 ELISA |
|
MEK1 ELISA |
|
c-Met ELISA |
|
Osteocalcin ELISA Human |
|
p38 MAPK ELISA |
|
p53 ELISA, Human |
|
Retinoblastoma ELISA, Human |
|
Src ELISA Human |
|
STAT1 ELISA Human |
|
Tau ELISA Murine (Tau ELISA murine 96) |
|
Tau ELISA Murine (Phospho-Tau [pSer199]) |
|
sTNF-RI ELISA Human |
|
sTNF RII ELISA Human |
|
TRAIL ELISA Human |
|
Solid phase sandwich Enzyme Linked-Immuno-Sorbent Assays (ELISA) employ an antibody (capture) specific for a protein (regardless of phosphorylation state) coated onto the wells of the multiwell plate. Samples, standards, control specimens and unknowns are incubated in the wells and antigen binds to the capture antibody. A second antibody specific for the protein in the assay (detection) is added and during the second one hour incubation, it binds to the immobilized protein captured during the first incubation. An anti-rabbit IgG-HRP completes the four-member sandwich. The reaction is visualized by the TMB substrate solution, which produces color, whose intensity is directly proportional to the concentration of the protein present in the original specimen. The reaction is stopped with a stop solution and read at 450 nm in the microtiter plate.
Features and Benefits Specific
- Specifically measures protein in questions with no crossreaction with other proteins
Sensitive
- Equal to or exceeds immunoblotting sensitivity
Convenient
- Kits contain all reagents needed for assay
- Precoated plates eliminate the coating step
- All incubations proceed at room temperature
- Any commercially available plate reader may be used
Accurate
- Control for total protein content with a pan protein ELISA
Quantitative
- Quantitate protein phosphorylation on a specific amino acid residue using a 96-well format
QC
- Lot-to-lot consistency of <10%
Validated
- Validated by peptide competition and mutant analysis ELISAs
Specificity
- Validated by peptide blocking and comparison with immunoblotting
back to top
FAM-FLICA Caspase Assays
Product Name |
Product Code
|
Caspase 1 Fluorescein (FLICA) Assay |
|
Caspase 2 Fluorescein (FLICA) Assay |
|
Caspase 9 Fluoroscein (FLICA) Assay |
|
Caspase 10 Fluorescein (FLICA) Assay |
|
Caspase 13 Fluorescein (FLICA) Assay |
|
Fluorometric or flow cytometry detection of active cellular caspases using Fluorochrome Inhibitor of Caspase activity (FLICA).
Features and Benefits
- Easy, fast and more convenient
- Caspases are earlier indicators of apoptosis than Tunnel or Annexin V
- Potent, cell-permeable and non-toxic fluorochrome inhibitor
- Specific for selected caspase based on inhibitor specificity
- A direct measure of apoptosis expressed as the number of active caspase enzymes present in the cell
- No need for cell lysis nor membrane permeabilization
- Kit could be run on fluorescence microscopes, fluorescence plate readers and flow cytometers
- Works on majority of species because caspases are well conserved through the species
back to top
Caspase Activity Assays
Product Name |
Product Code
|
Caspase 3 Assay Kit, Colorimetric |
|
Caspase 3 Assay Kit, Fluorimetric |
|
Caspase 6 Assay Kit, Colorimetric |
|
Caspase 6 Assay Kit, Fluorimetric |
|
Caspase 8 Assay Kit, Colorimetric |
|
Caspase 8 Assay Kit, Fluorimetric |
|
Fluorometric or flow cytometry detection of active cellular caspases using Fluorochrome Inhibitor of Caspase activity (FLICA).
Features and Benefits
- Easy, fast and more convenient
- Caspases are earlier indicators of apoptosis than Tunnel or Annexin V
- Potent, cell-permeable and non-toxic fluorochrome inhibitor
- Specific for selected caspase based on inhibitor specificity
- A direct measure of apoptosis expressed as the number of active caspase enzymes present in the cell
- No need for cell lysis nor membrane permeabilization
- Kit could be run on fluorescence microscopes, fluorescence plate readers and flow cytometers
- Works on majority of species because caspases are well conserved through the species
back to top
Fluorometric Cell-Associated Nitric Oxide Synthase (NOS) Detection System
Product Name |
Product Code
|
Nitric Oxide Synthase Detection System, Fluorimetric |
|
Sigma-RBI’s Fluorometric Cell-Associated Nitric Oxide Synthase Detection System is a simple and specific fluorometric assay for the measurement of free NO and NOS activity in living cells under physiological conditions. The need for radioisotope detection is replaced by the use of a cell-permeable diacetate derivative of the novel compound, 4,5-diaminofluorescein (DAF2-DA). The DAF2-DA penetrates cells rapidly and becomes hydrolyzed by an intracellular esterase to the parent DAF-2, which subsequently reacts with NO, produced by NOS, to form triazolofluorescein (DAF2-T). The fluorescent final product can then be quantified using an excitation filter at 492 nm and an emissions filter at 515 nm.
|
 |
Fluorescence detection of NO production in control and in LPS/IFN-γ-induced Raw 264.7 murine macrophage cells cultured in 96-well plates. Fluorescence signals were obtained after an incubation at room temperature for 110 min. Results were calculated as the percentage of the maximal signals recorded with 1 mM arginine and 2.5 µM NADPH in induced cultures. DPI: Diphenyleneiodonium (iNOS selective inhibitor).
Features and Benefits
- Non-radioactive, cell based
- Uses cell-permeable fluorescein to detect intracellular NOS
- Kinetic quantitative reaction in a spectrofluorometer, linear for 2 hours at room temperature
- Specific - requires induction of iNOS activity and inhibition of fluoresence generation by NOS inhibitors
- Convertible to high throughput screening for drug discovery efforts
back to top
Hydrogen Peroxide (H2O2) Colorimetric Assay
Product Name |
Product Code
|
Hydrogen Peroxide Colorimetric Assay (H2O2Colorimetric Assay) |
|
Measurement of H2O2 helps to determine how oxidative stress modulates various intracellular pathways. The H2O2 kit is designed to measure low concentrations of H2O2 in biological matrices. A color reagent produces a purple color proportional to the concentration of H2O2 in the samples or standards. The mechanism of the color reaction probably involves coordinated iron reacting with H2O2 and the dye molecule. The color reaction is read at 540-570 nm in the microplate spectrophotometer. Plot absorbance versus standard H2O2 concentrations to quantitate H2O2 in the samples.
Samples
- A wide range of samples may be used and diluted with 50 mM phosphate, pH 6.0 or cell culture media
Features and Benefits Simple
- No sample extraction or lysis required
- One step procedure with one reagent
Sensitive
- Detects H2O2 at 51.25 ng/ml when diluted in buffer
Quantitative
- All results are quantitative
- Standard concentration curves are run in each assay
Very Fast
- Results available within 30 minutes
Performance Characteristics
Precision
- Intra assay variations 3-10%
- Inter-assay variations 1.7-5.7%
Linearity
- A dilution of hydrogen peroxide was run multiple times in same assay
- The obtained line has a slope of 0.9342 with CV of 0.9983
- Plasma, urine and serum samples diluted 1:64 gave 90.6-105.6% recovery
Recovery
- Plasma, urine and serum samples spiked with H2O2 and diluted 1:64 gave >90% recovery
back to top
Kinase Assays –p38, JNK, MEK
Product Name |
Product Code
|
JNK 1&2 Activity Assay Kit |
|
p38 MAPK Activity Assay Kit |
|
MEK Activity Assay Kit |
|
Assays measure kinase phosphorylation activity, activation and inhibition by immunoblot detection. The endogenous kinase of interest is immunoprecipitated using the kinase specific antibody and EZview Red Protein A. The immunoprecipitated kinase phosphorylates a specific substrate. The kinase activity is then detected by an immunoblot analysis of the phosphorylated substrate using phosphospecific antibodies.
Features and Benefits
- A non-radioactive sensitive assay
- Optional radioactive protocol
- Contains all required reagents for the immunoprecipitation and phosphorylation reactions and also a detection antibody
- Easy and convenient procedure
- Friendly to use due to its components
- The EZview Red Affinity Gel: allows a better visualization of the precipitated fraction
- HRP conjugated detection antibody (JNK and p38 MAPK Activity Assay Kits component): no requirement for a secondary antibody, thus, reducing background activity
- p38 MAPK and MEK Activity Assay Kits contain a specific inhibitor for further verification of the kinase specificity
- Procedure is relatively short
- Reproducible results
- Useful for
- Studying kinases pathways
- Studying kinases, activators and inhibitors
- Studying and identifying extracellular stress stimuli that are converted into specific cellular responses through the activation of the specific kinase signaling pathway
back to top
Mitochondria Isolation Kit
Product Name |
Product Code
|
Mitochondria Isolation Kit |
|
Cytochrome c Oxidase Assay Kit |
|
The Mitochondria isolation kit enables the fast and easy isolation of an enriched mitochondrial fraction from animal tissues as well as the testing of the electrochemical proton gradient of the inner mitochondria-mediated apoptosis.
Features and Benefits
- Measures integrity of inner mitochondrial membrane (using fluorescent carbocyanine dye JC-1 supplied in the kit)
- Useful for isolating mitochondrial proteins for proteasome studies
- Can use Sigma’RBI’s Cytochrome c Oxidase Assay kit to measure outer membrane activity
- Suitable for 50 assays
back to top
Phosphospecific ELISAs
Product Name |
Product Code
|
Phospho-Akt/PKB [pThr308] ELISA |
|
Phospho-Akt/PKB [pSer473] ELISA |
|
Phospho-ATF2 [pThr69/pThr71] ELISA |
|
Phospho-Bcl-2 [pSer70] ELISA |
|
Phospho-CREB [pSer133] ELISA |
|
Phospho-ERK 1&2 [pThr185/pTyr187] ELISA |
|
Phospho-Heat Shock Protein 27 [pSer82] ELISA human |
|
IκBα ELISA human |
|
Phospho-Insulin Receptor β ?Subunit [pTyr1158] ELISA |
|
Phospho-Insulin Receptor β ?Subunit [pTyr1162/1163] ELISA |
|
Phospho-JNK1&2 [pThr183/pTyr185] ELISA |
|
Phospho-p38 MAPK [pThr180/pTyr182] ELISA |
|
Phospho-MEK1 [pSer218/pSer222] ELISA |
|
Phospho-c-Met [pTyr1230/pTyr1234/pTyr1235] ELISA |
|
Phospho-p53 [pSer15] ELISA human |
|
Phospho-p53 [pSer15] ELISA human |
|
Phospho-Retinoblastoma [pSer249/pThr252] ELISA, human |
|
Phospho-Retinoblastoma [pThr821] ELISA, human |
|
Phospho-Src [pTyr418] ELISA, human |
|
Phospho-STAT1 [pTyr701] ELISA |
|
Phospho-Tau [pSer199] ELISA, murine |
|
Phosphospecific ELISAs offer a sensitive, quantitative and economical alternative to traditional immunoblotting or functional assays.
The format is a solid phase sandwich Enzyme-Linked Immunosorbent Assay (ELISA). A capture antibody specific for a protein (regardless of phosphorylation state) has been coated onto the wells of a multiwell plate. Antigen binds to the capture antibody. A detection antibody specific for the phosphorylated or nonphosphorylated protein is added and binds to the immobilized protein. An anti-rabbit IgG-HRP completes the four member sandwich. The reaction is visualized by the tetramethylbenzidene (TMB) substrate and the intensity of the color is directly proportional to the concentration of protein present in the original sample. The reaction is read at 340 nm in a multiwell plate reader.
Samples
Cell extracts are the samples of choice, because the proteins involved in the phosphorylation are present in intracellular compartments. Researchers may use the cell extraction procedure provided in the technical bulletin or utilize the Sigma Mammalian Cell Lysis Kit.
Features and Benefits Sensitive
- picograms/ml quantities for non-phosphospecific ELISAs
- below 1.0 unit/ml for phoshospecific kits
- 2 to 10 times more sensitive than traditional immunoblots
Sensitive
- Highly specific for target protein and phosphorylation site
- Validated against immunoblotting performed with the same antigen and antibodies
- Validated by peptide competition
Quantitative
- All results are quantitative
- Standard concentration curve is run in each assay
Simple
- Precoated plates eliminate the coating step
- All incubations at room temperature
- Small sample size – only 10 µl (diluted to a total volume of 100 µl)
- Use any multiwell reader available on the market
- No additional equipment required
Stable
- Entire kit is stable in the refrigerator for 12 months
- Components are all stored at the same temperature
- Liquid stable chromogen and stop reagents
- Two vials of standard extend the kit life span
Fast
Economical
- Reagents may be used for multiple kit runs
Performance Characteristics
- Intra and Inter-assay variations
- Linearity of dilution
- Recovery – ranges from 93 to 97%
- Each phosphospecific kit is normalized against non-phosphospecific ELISA
 Sample Data Using Sigma-RBI Phosphospecific ELISAs
Detection of p38 MAPK [pThr180/pTyr182] by ELISA vs. Immunoblotting. The sensitivity of this ELISA was compared to immunoblotting using known quantities of p38 MAPK [pThr180/pTyr182]. The data presented show that the sensitivity of the ELISA is approximately 10X greater than that of immunoblotting. The bands shown in the immunoblotting data were developed using rabbit anti-p38 MAPK [pThr180/pTyr182], an alkaline phosphatase conjugated anti-rabbit IgG, followed by chemiluminescent substrate and autoradiography.
Recombinant p38 MAPK was phosphorylated using MKK6 enzyme in vitro. Non-phosphorylated p38 MAPK was used as control. The phosphorylated and non-phosphorylated p38 MAPK were analyzed with phospho-p38 MAPK [pThr180/pTyr182] ELISA and p38 MAPK ELISA. This phospho-p38 MAPK (pThr180/pTyr182) ELISA kit is specific for measurement of phosphorylated p38 MAPK at threonine 180 and tyrosine 182. The kit does not detect non-phosphorylated p38 MAPK protein.
back to top
Glutathione Assay Kit: For the measurement of total glutathione in biological samples
Product Name |
Product Code
|
Glutathione Assay Kit |
|
Reduced glutathione (GSH), a tripeptide (γ-glutamyl-cysteinylglycine), is involved in many biological processes such as detoxification of xenobiotics, removal of hydroperoxides, and maintenance of the oxidation state of protein sulfhydryls. It is the key antioxidant present in animal tissues.
The kit provides all reagents for the simple and quick assay of the level of total glutathione (oxidized glutathione and GSH) in a cell, tissue extracts, red blood cells or plasma.
The biological samples are first deproteinized using 5% sulfosalicylic acid solution. The total glutathione content of the sample is then assayed using a kinetic assay in which catalytic amounts of glutathione cause a continuous reduction of 5,5’-dithiobis-(2-nitrobenzoic acid (DTNB) to TNB. The oxidized glutathione formed is recycled by glutathione reductase and NADPH. The product, TNB, is assayed colorimetrically at 412 nm.
Assaying in a 96 well plate maximizes the number of samples that can be tested.
Features and Benefits
- picograms/ml quantities for non-phosphospecific ELISAs
- below 1.0 unit/ml for phoshospecific kits
- 2 to 10 times more sensitive than traditional immunoblots
Sensitive
- A quick and simple method
- The kit provides all the reagents required for a colorimetric assay of total glutathione (oxidized + GSH)
- The kit includes a deproteination reagent for biological samples
Standard curve of reduced glutathione as measured in the kinetic colorimetric assay:
back to top
Glutathione S-Transferase (GST) Assay Kit: For the measurement of GST in biological samples
Product Name |
Product Code
|
Glutathione-S-Transferase (GST) Assay Kit |
|
Glutathione S-transferases (GSTs) are a group of enzymes important in the detoxication of many xenobiotics in mammals. The enzymes protect cells against toxicants by conjugating the thiol group of the glutathione to electrophilic xenobiotics, and thereby defend cells against the mutagenic, carcinogenic, and toxic effects of the compounds. GST activity is present in plants, insects, yeast, bacteria, and in most mammalian tissues, especially in the liver, which plays a key role in detoxification.
Glutathione S-Transferase (GST) Assay Kit assays total GST activity by measuring the conjugation of 1-chloro-2,4-dinitrobenzene (CDNB) with reduced glutathione. Product formation (GS-DNB conjugate) is detected at 340 nm, which makes possible a spectrophotometric assay for GST activity. The rate of increase of absorption is directly proportional to the GST activity in the sample.
Features and Benefits
- All the reagents required for a fast and easy measurement of total GST activity
- A GST positive control
- Used to measure GST activity in cell and bacterial lysates, tissue homogenates and in plasma and erythrocyte lysates
- Sufficient for 100 assays in 1 ml cuvettes or 500 reactions in 96 well plates
Measurement of Glutathione-S-Transferase (GST) activity:
The activity of GST enzyme was measured at different concentrations in a 1 ml cuvette using the GST Assay Kit. The graph demonstrates a linear ratio between the amount of enzyme (over the range ~0.05-0.5 µg) and the enzyme activity, as measured by the increase in OD340 units per min.
back to top
Casein Kinase Kits
The three casein kinase kits provide an easy method for measuring the activity of the different casein kinases by an in vitro phosphorylation of casein substrates. The kits can be used with cell lysates, tissue homogenates, column fractions, or the purified enzyme.
Casein kinases (CK) are highly conserved serine/threonine kinases, ubiquitous in eukaryotic organisms, and defined by their substrate preference for casein. They are insensitive to cyclic nucleotides, calcium, and phospholipids. Casein kinases are grouped into two main families, type I and type II, according to their structure, specificity and response to effectors.
Features and Benefits
- Easy method for measuring the activity of the different casein kinases by in vitro phosphorylation of substrate
- Includes specific inhibitors to allow the distinction between the CK activities
- Can be used for CK activity measurement in cell lysates, tissue homogenates, column fractions or a purified enzyme
Casein Kinase Assay Kit
The Casein Kinase Assay Kit (general) measures the activity of the different casein kinases by an in vitro phosphorylation of α-casein. The a-casein substrate is phosphorylated by both CKI and CKII. The kits includes CKI and CKII specific inhibitors to distinguish between CKI and CKII activities in the biological sample.
Product Name |
Product Code
|
Casein Kinase Assay Kit |
|
Casein Kinase I Assay Kit
The Casein Kinase I (CKI) Assay Kit measures the activity of Casein Kinase I by an in vitro phosphorylation of a Casein Kinase I phosphopeptide substrate (a specific CKI substrate). In addition, the kit contains a CKI specific inhibitor (IC261) to ensure the specificity of the CK activity measured and a purified CKId enzyme to serve as a positive control.
Product Name |
Product Code
|
Casein Kinase II (CKII) Assay Kit |
|
Casein Kinase II Assay Kit
The Casein Kinase II Assay Kit measures the CKII enzyme activity by an in vitro phosphorylation of the Casein Kinase II Substrate. This substrate is a peptide that is selective for CKII with not activity with CKI. The kit also contains a CKI specific inhibitor to ensure the specificity of the CK activity and in addition, it also includes purified CKII for use as a positive control.
Product Name |
Product Code
|
Casein Kinase II (CKII) Assay Kit |
|
back to top
DHFR Assay Kit: For the detection of Dihydrofolate Reductase (DHFR) activity and for screening of DHFR inhibitors
Product Name |
Product Code
|
Dihydrofolate Reductase Assay Kit |
|
DHFR is an enzyme catalyzing a reaction essential for the biosynthesis of nucleotidic bases of DNA. Blockade of the DHFR enzyme causes cell death as a result of inhibition of DNA synthesis. For this reason, DHFR is considered a target for the development of anti-tumor drugs.
The assay is based on the ability of DHFR to catalyze the reversible NADPH-dependent reduction of dihydrofolic acid to tetrahydrofolic acid. The reaction progress is followed by monitoring the decrease in absorbance at 340 nm.
|
DHFR |
|
Dihydrofolic acid + NADPH + H+ |
—> <— |
Tetrahydrofolic acid + NADP+ |
Features and Benefits
- Contains all the reagents required for a colorimetric assay of DHFR activity in cell lysates, tissue homogenates, or column fractions of purified enzyme
- Includes a purified enzyme to be used as a positive control and for screening of DHFR inhibitors
- Includes methotrexate (MTX), a prokaryotic and eukaryotic DHFR specific inhibitor, which exhibits anti-tumor activities
- Tested using A431, NIH 3T3 and CHO cell lines, rat liver, kidney, brain, and skeletal muscle tissue extracts and recombinant DHFR
- Sufficient reagents are supplied for 50-100 1ml tests
Activity of recombinant Dihydrofolate Reductase (DHFR) in the presence of the inhibitor methotrexate (MTX). Specific activity (Unit/mg protein) of recombinant DHFR was measured in the absence or in the presence of different concentrations of MTX. 50% inhibition of the enzyme is observed at a concentration of 0.025 µM MTX.
back to top
New Retinoblastoma Phospho-Specific ELISAs and Antibodies
Product Name |
Product Code
|
Retinoblastoma ELISA, Human |
|
Phospho-Retinoblastoma [pSer249/pThr252] ELISA, Human |
|
Phospho-Retinoblastoma [pThr821] ELISA, Human |
|
Features and Benefits
- Very low intra-assay variability from 2.84% to 7.58%
- Very low inter-assay variability from 5.7% to 9.21%
- Linearity of dilution – correlation coefficient for various dilutions was 0.99 for both assays
- Recover – average recovery: 92.7% and 10% respectively
- Suitable for 96 wells
Retinoblastoma protein (Rb), the tumor suppressor product of the retinoblastoma susceptibility gene, is a 110 kDa protein that functions as a negative regulator of the cell cycle by arresting cells in the G1 phase and halting inappropriate cell proliferation. Loss of Rb function leads to uncontrolled cell growth and tumor development and is found in all retinoblastomas and in a variety of other human malignancies including cancers of the breast, lung, colon, prostate, osteosarcomas, soft tissue sarcomas and leukemia. The ability of Rb protein to alter transcription is regulated by phosphorylation. Rb contains at least 16 consensus sequences for cdk phosphorylation. The dephosphorylation of the RB protein returns Rb to its active, growth suppressive state.
|
![Peptide Competition with Anti-phospho Rb [pThr826]](/content/dam/sigma-aldrich/life-science/cell-signaling-and/Highlights08.gif) Peptide Competition with Anti-phospho Rb [pThr826]
|
Human Rb ELISA is designed for in vitro quantitative detection of non-phosphorylated, phosphorylated and dually phosphorylated human Rb in cell lysates. A monoclonal capture antibody specific for Rb protein (regardless of phosphorylation state) has been coated onto a 96 multiwell plate. Rb antigen in samples and standards binds to the capture antibody. A detection antibody, specific for the phosphorylated or non-phosphorylated human Rb, is added and binds to the immobilized Rb protein. A streptavidin-HRP or anti-rabbit IgG-HRP completes the four-membrane sandwich. The reaction is visualized by a TMB substrate and the intensity of the color is directly proportional to the concentration of Rb present in the original sample. The reaction is read at 450 nm in a multiwell plate reader. The assays detect both native and recombinant human Rb and phospho-Rb [pSer249/pThr252] and pThr821] proteins. The assays are highly sensitive (<0.1 ng/mL and <0.8 units/mL, respectively), specific fast (4 hours total time) and simple (offer precoated plates, all incubations proceed at room temperature and the reagents are supplied ready to use).