Cell Biology
PRECISIO® Kinases
Our PRECISIO Kinases ensure the quality and lot-to-lot consistency that your research demands. Our large portfolio of active kinases exhibit high purity and specific activity, and each PRECISIO Kinase is tested using [32P] radioactive assay for quality activity results. Detailed product datasheets containing optimized assay protocols allow you to use PRECISIO Kinases with confidence.
We offer a broad assortment of both fragments and full-length protein kinases suitable for kinase assays, western blotting, high throughput screening and kinase profiling. PRECISIO Kinases are complemented by our vast collection of inhibitors and activators, antibodies, substrates, and assay kits.
See our Active Kinases A–Z or search using our Kinase Search box below.
Over 350 Active Kinases in Stock
  • Cost effective 10 µg and BULK quantities
  • Complimentary inhibitors, activators, substrates and antibodies
Use with Confidence
  • Optimized assay protocols provided
  • High purity and specific activity
  • Lot-to-lot consistency
  • Suitable for high throughput screening and kinase profiling
Representative Quality Data
Specific Activity of CHK1 Active Image
Figure 1: Specific Activity of CHK1 Active (Product No. C0870) 187 nmol/min/mg (pmole/min/µg)
SDS-PAGE Gel of CHK1 Active Image
Figure 2: SDS-PAGE Gel of CHK1 Active (Product No. C0870) >95% by densitometry