HL-1 Cardiac Muscle Cell Line
Discovered by Dr. William Claycomb, HL-1 cells are the first cell line established that can maintain the differentiated cardiomyocyte phenotype and contractile activity in vitro. The HL-1 cell line can be used for the study of cardiac cell hypertrophy that follows myocardial infarction, the testing of novel cardiotherapeutic drugs and treatments, the production of high levels of cardiac proteins and the study of mature cardiomyocyte specific genes.
 Featured Products & Resources
Sera: HL-1 qualified
This FBS has been qualified using HL-1 mouse cardiac muscle cells
Ordering Information