| Antibiotics/Antimycotics |
| Life Technologies/Gibco Product No. |
Sigma Product No. |
Product Description |
Product Differences |
|---|
| 15240062 |
A5955 |
Antibiotic Antimycotic Solution, stabilized (100X) |
Exact match. No differences. |
| 15240096 |
A5955 |
Antibiotic Antimycotic Solution, stabilized (100X) |
Exact match. No differences. |
| 10131035 |
G8168 |
G418 disulfate solution |
Exact match. No differences. |
| 11811023 |
A1720 |
Geneticin disulfate salt |
Sigma product is non-branded form of Geneticin. |
| 11811031 |
A1720 |
Geneticin disulfate salt |
Sigma product is non-branded form of Geneticin. |
| 11811098 |
A1720 |
Geneticin disulfate salt |
Sigma product is non-branded form of Geneticin. |
| 15710049 |
G1272 |
Gentamicin solution |
Exact match. No differences. |
| 15750045 |
G1397 |
Gentamicin solution |
Exact match. No differences. |
| 10687010 |
H0654 |
Hygromycin B solution |
Sigma is powder, Life Technologies/Gibco is liquid. |
| 15160047 |
K0129 |
Kanamycin solution from Streptomyces Kanamyceticus |
Exact match. No differences. |
| 10378016 |
G1146 |
L-Glutamine-Penicillin-Streptomycin solution |
Sigma product is in 0.9% NaCl. Life Technologies/ Gibco is in 10mM citrate buffer. |
| 15640055 |
P4083 |
Penicillin - Streptomycin - Neomycin solution stabilized (100X) |
Exact match. No differences. |
| 15140122 |
P4333 |
Penicillin - Streptomycin Solution (100X) Stabilized |
Exact match. No differences. |
| 15140148 |
P4333 |
Penicillin - Streptomycin Solution (100X) Stabilized |
Exact match. No differences. |
| 15070063 |
P4458 |
Penicillin - Streptomycin Solution (50X) Stabilized |
Exact match. No differences. |
| |
| Serum |
| Life Technologies/Gibco Product No. |
Sigma Product No. |
Product Description |
Product Differences
|
|---|
| 16170078 |
12133C |
Calf Serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 16170086 |
12133C |
Calf Serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 16110082 |
C5405 |
Chicken Serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 10082139 |
F4135 |
Fetal Bovine Serum |
Exact match. No differences. |
| 10082147 |
F4135 |
Fetal Bovine Serum |
Exact match. No differences. |
| 10106151 |
F7524 |
Fetal Bovine Serum |
Exact match. No differences. |
| 10106169 |
F7524 |
Fetal Bovine Serum |
Exact match. No differences. |
| 16000036 |
F2442 |
Fetal Bovine Serum |
Exact match. No differences. |
| 16000044 |
F2442 |
Fetal Bovine Serum |
Exact match. No differences. |
| 26140079 |
F2442 |
Fetal Bovine Serum |
Recommended higher quality alternative. |
| 26140087 |
F2442 |
Fetal Bovine Serum |
Recommended higher quality alternative. |
| 26400036 |
F0392 |
Fetal Bovine Serum, Dialyzed |
Exact match. No differences. |
| 26400044 |
F0392 |
Fetal Bovine Serum, Dialyzed |
Exact match. No differences. |
| 10108157 |
F9665 |
Fetal Bovine Serum, Heat Inactivated |
Exact match. No differences. |
| 10108165 |
F9665 |
Fetal Bovine Serum, Heat Inactivated |
Exact match. No differences. |
| 16140063 |
F4135 |
Fetal Bovine Serum, Heat Inactivated |
Recommended higher quality alternative. |
| 16140071 |
F4135 |
Fetal Bovine Serum, Heat Inactivated |
Recommended higher quality alternative. |
| 16210064 |
G6767 |
Goat Serum Donor Herd |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 16210072 |
G6767 |
Goat Serum Donor Herd |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 16050122 |
H1270 |
Horse Serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 16050130 |
H1270 |
Horse Serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 26050070 |
H1138 |
Horse Serum, Heat Inactivated |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 26050088 |
H1138 |
Horse Serum, Heat Inactivated |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 16010159 |
N4637 |
Newborn Calf Serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 16010167 |
N4637 |
Newborn Calf Serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 26010066 |
N4762 |
Newborn Calf Serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 26010074 |
N4762 |
Newborn Calf Serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 26250084 |
P9783 |
Porcine Serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand origin |
| 16120099 |
R4505 |
Rabbit serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand or US origin |
| 16120107 |
R4505 |
Rabbit serum |
Sigma is US origin, Life Technologies/Gibco is New Zealand or US origin |
| 16070096 |
S2263 |
Sheep Serum |
Sigma is adult animal of US origin, Life Technologies/Gibco is lamb of New Zealand origin |
| |
| Media Supplements |
| Life Technologies/Gibco Product No. |
Sigma Product No. |
Product Description |
Product Differences
|
|---|
| 15630049 |
H0887 |
HEPES solution |
Exact match. No differences. |
| 15630056 |
H0887 |
HEPES solution |
Exact match. No differences. |
| 41065012 |
H0137 |
HT Media Supplement (50X) Hybri-Max. |
Closest Alternative. Sigma product is a power. Life Technologies/ Gibco product is a liquid. |
| 41400045 |
I3146 |
ITS Liquid Media Supplement (100X) |
Exact match. No differences. |
| 25030024 |
G7513 |
L-Glutamine solution |
Sigma’s glutamine is in H20, Life Technologies glutamine is in NaCl |
| 25030032 |
G7513 |
L-Glutamine solution |
Sigma’s glutamine is in H20, Life Technologies glutamine is in NaCl |
| 11130036 |
M5550 |
MEM Amino Acids Solution (50X) |
Exact match. No differences. |
| 11140035 |
M7145 |
MEM Non-essential Amino Acid Solution (100X) |
Exact match. No differences. |
| 11140068 |
M7145 |
MEM Non-essential Amino Acid Solution (100W) |
Exact match. No differences. |
| 11120037 |
M6895 |
MEM Vitamin Solution (100W) |
Exact match. No differences. |
| 51500056 |
S4920 |
SITE Liquid Media Supplement (100X) |
Exact match. No differences. |
| 11360039 |
S8636 |
Sodium pyruvate solution |
Exact match. No differences. |
| |
| Balanced Salt Solutions |
| Life Technologies/Gibco Product No. |
Sigma Product No. |
Product Description |
Product Differences
|
|---|
| 14040083 |
D8662 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14040091 |
D8662 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14040117 |
D8662 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14040174 |
D8662 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14080048 |
D1283 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14190094 |
D8537 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14190136 |
D8537 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14190169 |
D8537 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14200067 |
D1408 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14155048 |
E3024 |
Earle's Balanced Salt Solution |
Sigma product E3024 (no phenol red) or E6267 (no calcium or magnesium). |
| 14155048 |
E6267 |
Earle's Balanced Salt Solution |
Sigma product E3024 (no phenol red) or E6267 (no calcium or magnesium). |
| 14025050 |
H8264 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14065049 |
H1641 |
Hanks' Balanced Salt Solutions |
Sigma contains phenol red, Life Technologies/Gibco is without phenol red |
| 14170070 |
H9394 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14170088 |
H9394 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14170138 |
H9394 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14175053 |
H6648 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14185045 |
H4641 |
Hanks' Balanced Salt Solutions |
Sigma contains phenol red, Life Technologies/Gibco is without phenol red |
| 24020083 |
H9269 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 24020091 |
H9269 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 20012019 |
806544 |
Phosphate buffer pH 7.2 at 25° C |
Exact match. No differences. |
| 70013016 |
806552 |
Phosphate buffer pH 7.2 at 25° C |
Exact match. No differences. |
| 70011036 |
806552 |
Phosphate Buffered Saline, pH 7.4 |
Exact match. No differences. |
| 10010015 |
806552 |
Phosphate Buffered Saline, pH 7.4 |
Exact match. No differences. |
| |
| Classical Media |
| Life Technologies/Gibco Product No. |
Sigma Product No. |
Product Description |
Product Differences
|
|---|
| 41010026 |
B1522 |
Basal Medium Eagle |
Exact match. No differences. |
| 10938025 |
D5671 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 21013024 |
D0422 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 31053044 |
D1145 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 21969035 |
D6546 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 31053028 |
D1145 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 31966021 |
D0822 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 32430027 |
D0819 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 41965039 |
D5796 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 41965047 |
D5796 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 41965062 |
D5796 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 41965120 |
D5796 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 41966029 |
D6429 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 41966052 |
D6429 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 42430025 |
D4822 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 12800017 |
D5648 |
Dulbecco's Modified Eagle's Medium - high glucose |
Closest alternative. Sigma product does not contain sodium pyruvate. Invitrogen product contains sodium pyruvate. |
| 52100039 |
D5648 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 61965026 |
D0819 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 31885023 |
D6046 |
Dulbecco's Modified Eagle's Medium - low glucose |
Exact match. No differences. |
| 31885049 |
D6046 |
Dulbecco's Modified Eagle's Medium - low glucose |
Exact match. No differences. |
| 11039021 |
D6434 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Sigma is without L-glutamine, Life Technologies/Gibco contains L-glutamine |
| 21041025 |
D0547 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Closest alternative. Sigma product contains phenol red. Invitrogen product does not contain phenol red. |
| 21331020 |
D6421 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Contains HEPES. Differences in L-valine, CaCl2•2H2O and MgCl2 concentrations |
| 31330038 |
D8437 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Exact match. No differences. |
| 31331028 |
D0697 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Sigma product contains HEPES, Life Tech product does not contain HEPES. |
| 32500043 |
D0547 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Exact match. No differences. |
| 42400010 |
D8900 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Exact match. No differences. |
| 21300017 |
D5773 |
Dulbecco's Phosphate Buffered Saline |
Sigma's does not contain CaCl2, Life Technologies/Gibco supplies a bottle of CaCl2. |
| 21300058 |
D5773 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 21710025 |
G2549 |
Glasgow Minimum Essential Medium |
Exact match. No differences. |
| 31980022 |
I2911 |
Iscove's Modified Dulbecco's Medium |
Exact match. No differences. |
| 42200014 |
I7633 |
Iscove's Modified Dulbecco's Medium |
Exact match. No differences. |
| 42200030 |
I7633 |
Iscove's Modified Dulbecco's Medium |
Exact match. No differences. |
| 11415049 |
L5520 |
L-15 Medium (Leibovitz) |
Sigma is without L-glutamine, Life Technologies/Gibco contains L-glutamine |
| 21083027 |
L1518 |
L-15 Medium (Leibovitz) |
Sigma product contains phenol red. Invitrogen product does not contain phenol red. |
| 41300021 |
L4386 |
L-15 Medium (Leibovitz) |
Exact match. No differences. |
| 41300070 |
L4386 |
L-15 Medium (Leibovitz) |
Exact match. No differences. |
| 22330021 |
M9309 |
McCoy's 5A Medium |
Sigma product is without HEPES. Invitrogen product contains 25 mM HEPES |
| 26600023 |
M8403 |
McCoy's 5A Medium |
Sigma is without L-glutamine, Life Technologies/Gibco contains L-glutamine |
| 22340020 |
M7528 |
Medium 199 |
Sigma is without L-glutamine, Life Technologies/Gibco contains L-glutamine |
| 31100019 |
M5017 |
Medium 199 |
Exact match. No differences. |
| 31100027 |
M5017 |
Medium 199 |
Exact match. No differences. |
| 31150022 |
M4530 |
Medium 199 |
Exact match. No differences. |
| 31985047 |
M3817 |
MegaCell™ RPMI-1640 Medium |
Recommended alternative. |
| 31985054 |
M3817 |
MegaCell™ RPMI-1640 Medium |
Recommended alternative. |
| 51985026 |
M3817 |
MegaCell™ RPMI-1640 Medium |
Closest alternative. Sigma is without L-glutamine, Invitrogen contains dipeptide Glutamax. |
| 11700077 |
M0769 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 11900016 |
M0644 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 11900073 |
M0644 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 12000014 |
M0894 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 12000063 |
M0894 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 21090022 |
M2279 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 21090055 |
M2279 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 22561021 |
M0200 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 31095029 |
M4655 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 31095052 |
M4655 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 32360026 |
M7278 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 32561029 |
M6199 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 32571028 |
M6074 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 41061029 |
M0450 |
Minimum Essential Medium Eagle |
Sigma contains phenol red, Life Technologies/Gibco is without phenol red |
| 41090028 |
M0446 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 41500018 |
M0643 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 41500067 |
M0643 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 41500083 |
M0643 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 42360024 |
M7278 |
Minimum Essential Medium Eagle |
Sigma is without L-glutamine, Life Technologies/Gibco contains dipeptide Glutamax |
| 61100053 |
M0268 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 61100087 |
M0268 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 61100103 |
M0268 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 21340039 |
N1140 |
NCTC 109 Medium |
Exact match. No differences. |
| 22390025 |
N2147 |
Nutrient Mixture F-10 Ham |
Sigma is without L-glutamine, HEPES concentration is different |
| 31550023 |
N6908 |
Nutrient Mixture F-10 Ham |
Exact match. No differences. |
| 21127022 |
N3520 |
Nutrient Mixture F-12 Ham |
Sigma is powder, Life Technologies/Gibco is liquid |
| 21700018 |
N6760 |
Nutrient Mixture F-12 Ham |
Exact match. No differences. |
| 21700026 |
N6760 |
Nutrient Mixture F-12 Ham |
Exact match. No differences. |
| 21765029 |
N6658 |
Nutrient Mixture F-12 Ham |
Exact match. No differences. |
| 21765037 |
N6658 |
Nutrient Mixture F-12 Ham |
Exact match. No differences. |
| 31765027 |
N3790 |
Nutrient Mixture F-12 Ham |
Exact match. No differences. |
| 27016021 |
R1145 |
RPMI 1640 Medium |
Closest alternative. Sigma product contains phenol red. Invitrogen product does not contain phenol red. |
| 61870010 |
R2405 |
RPMI 1640 Medium |
Exact match. No differences. |
| 13018015 |
R4130 |
RPMI-1640 Medium |
Exact match. No differences. |
| 13018031 |
R4130 |
RPMI-1640 Medium |
Exact match. No differences. |
| 52400017 |
R5158 |
RPMI-1640 Medium |
Exact match. No differences. |
| 52400025 |
R5158 |
RPMI-1640 Medium |
Exact match. No differences. |
| 52400041 |
R5158 |
RPMI-1640 Medium |
Exact match. No differences. |
| 72400021 |
R7388 |
RPMI-1640 Medium |
Closest alternative. Sigma product contains L-glutamine and does not contain sodium bicarbonate. Invitrogen product contains dipeptide Glutamax and sodium carbonate. |
| |
| Other Reagents |
| Life Technologies/Gibco Product No. |
Sigma Product No. |
Product Description |
Product Differences
|
|---|
| 31350010 |
M3148 |
2-Mercaptoethanol |
Sigma is pure reagent, Life Technologies/Gibco is 50mM solution |
| 35050038 |
G8541 |
Ala-Gln |
Exact match. No differences. |
| 15260037 |
A8412 |
Albumin solution from bovine serum |
Exact match. No differences. |
| 11108016 |
T1428 |
apo-Transferrin |
Exact match. No differences. |
| 10908010 |
B3795 |
Brain-derived Neurotrophic Factor human |
Exact match. No differences. |
| 12648010 |
C6164 |
Cell Freezing Medium-DMSO (1W) |
Recommended alternative. |
| 15210040 |
D1925 |
Colcemid. solution |
Exact match. No differences. |
| 15212012 |
D1925 |
Colcemid. solution |
Sigma in HBSS, Life Technologies/Gibco in PBS |
| 17018029 |
C9891 |
Collagenase from Clostridium histolyticum |
Recommended product, consult Certificate of Analysis for exact specifications. |
| 17104015 |
C5138 |
Collagenase from Clostridium histolyticum |
Recommended product, consult Certificate of Analysis for exact specifications. |
| 17105041 |
D4693 |
Dispase from Bacillus polymyxa |
Recommended product, consult Certificate of Analysis for exact units of activity. |
| PHG0315 |
E9644 |
Epidermal Growth Factor human |
Exact match. No differences. |
| 13241013 |
F5542 |
Fibroblast Growth Factor-Acidic human |
Exact match. No differences. |
| 13256029 |
F0291 |
Fibroblast Growth Factor-Basic human |
Exact match. No differences. |
| 33016015 |
F2006 |
Fibronectin from human plasma |
Exact match. No differences. |
| 10092013 |
G9641 |
Giemsa Solution |
Exact match. No differences. |
| PHC7045 |
G1777 |
Glial Cell Line-derived Neurotrophic Factor human |
Exact match. No differences. |
| 11107018 |
T1283 |
holo-Transferrin bovine |
Exact match. No differences. |
| 10598H24E50 |
I3769 |
Insulin-like Growth Factor-I human |
Exact match. No differences. |
| 23017015 |
L2020 |
Laminin from Engelbreth-Holm-Swarm murine sarcoma (basement membrane) |
Exact match. No differences. |
| 11905031 |
L0288 |
Lipid Mixture 1, Chemically Defined |
Exact match. No differences. |
| 13257019 |
N6009 |
Nerve Growth Factor-2.5S from murine submaxillary gland |
Exact match. No differences. |
| 13290010 |
N0513 |
Nerve Growth Factor-7S from murine submaxillary gland |
Exact match. No differences. |
| PHG0045 |
P3201 |
Platelet-Derived Growth Factor-BB human |
Exact match. No differences. |
| 24040032 |
P5556 |
Pluronic F-68 solution |
Exact match. No differences. |
| 10575090 |
P9327 |
Potassium chloride solution |
Exact match. No differences. |
| 25080060 |
S8761 |
Sodium bicarbonate solution |
Exact match. No differences. |
| 15250061 |
T8154 |
Trypan Blue solution |
Exact match. No differences. |
| 17075029 |
T6522 |
Trypsin inhibitor from Glycine max (soybean) |
Recommended alternative. |
| 27250018 |
T4799 |
Trypsin porcine pancreas |
Recommended alternative. |
| 25050014 |
T4424 |
Trypsin solution from porcine pancreas |
Exact match. No differences. |
| 25050030 |
T4424 |
Trypsin solution from porcine pancreas |
Exact match. No differences. |
| 15400054 |
T4174 |
Trypsin solution from porcine pancreas |
Exact match. No differences. |
| 15400054 |
T3924 |
Trypsin-EDTA solution |
Closest Alternative. Invitrogen product does not contain phenol red. Sigma product contains phenol red. |
| 25200056 |
T4049 |
Trypsin-EDTA solution |
Sigma product does not contain phenol red. Invitrogen product does contain phenol red. |
| 25200072 |
T4049 |
Trypsin-EDTA solution |
Sigma product does not contain phenol red. Invitrogen product does contain phenol red. |
| 25300054 |
T3924 |
Trypsin-EDTA solution |
Exact match. No differences. |
| 25300062 |
T3924 |
Trypsin-EDTA solution |
Exact match. No differences. |
| 15230071 |
W3500 |
Water |
Exact match. No differences. |
| 15230089 |
W3500 |
Water |
Exact match. No differences. |
| 15230097 |
W3500 |
Water |
Exact match. No differences. |
| 15230188 |
W3500 |
Water |
Exact match. No differences. |
| 18180059 |
Y1000 |
Yeast extract |
Sigma is powder, Life Technologies/Gibco is liquid |
| 18200048 |
Y4375 |
Yeast Extract Ultrafiltrate (50W) |
Exact match. No differences. |
| |
| Serum Free and Speciality Media |
| Life Technologies/Gibco Product No. |
Sigma Product No. |
Product Description |
Product Differences
|
|---|
| 10743011 |
C4726 |
CHO Medium |
Recommended alternative. |
| 10743029 |
C4726 |
CHO Medium |
Recommended alternative. |
| 12052098 |
C5467 |
CHO Medium |
Recommended alternative. Sigma product is without L-Glutamine. Invitrogen product is with L-Glutamine. |
| 12052114 |
C5467 |
CHO Medium |
Recommended alternative. Sigma product is without L-Glutamine. Invitrogen product is with L-Glutamine. |
| 31033020 |
C5467 |
CHO Medium |
Closest alternative. Sigma product does not contain L-Glutamine. Invitrogen product does not contain L-Glutamine. |
| 10744019 |
S0196 |
EpiLifed Keratinocyte Medium |
Exact match. No differences. |
| 11300027 |
G9771 |
Grace's Insect Medium |
Exact match. No differences. |
| 11279023 |
H4409 |
Hybridoma Medium, Animal Component-free |
Recommended alternative. |
| 12045076 |
H4281 |
Hybridoma Medium, Serum-free |
Recommended alternative. |
| 12045084 |
H4281 |
Hybridoma Medium, Serum-free |
Recommended alternative. |
| 11405057 |
I7760 |
IPL-41 Insect Medium |
Exact match. No differences. |
| 10372019 |
M8537 |
MCDB 131 Medium |
Sigma product is powder and contains L-Glutamine, Invitrogen product is liquid and does not contain L-Glutamine. |
| 11681020 |
M3678 |
MDCK Protein-free Medium |
Exact match. No differences. |
| 21720024 |
S0146 |
Schneider's Insect Medium |
Exact match. No differences. |
| 10744019 |
S0196 |
Stemline® Keratinocyte Medium II |
Exact match. No differences. |
| 10639011 |
S0189 |
Stemlined Hematopoietic Stem Cell Expansion Medium |
Recommended alternative. |