StableCell™ Trypsin Sample Request
Please complete this form to receive a free sample of our new StableCell™ Trypsin solution.* This modern formulation of trypsin is stable at room temperature, but performs identically to traditional trypsin solutions that must be stored frozen at -20°C to prevent autodigestion. StableCell™ Trypsin solution is specifically designed to perform cell detachment without the need to aliquot, freeze, and thaw the trypsin. This saves significant time by eliminating time-consuming aliquoting and waiting for trypsin to thaw for cell passaging or experiments.
For more information on StableCell™ Trypsin, visit this page.
Ready for your free sample? Thanks for filling out the form below.
*Free sample offer valid for US customers only at this time. For more information on sample availability in other countries, please contact your local sales rep.