Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Biological Buffers
ACES
ACES is a zwitterionic compound used in biochemical and biological research1. ACES has been utilized to develop buffers for both agarose and polyacrylamide gel electrophoresis2. In addition, this buffer is an ideal separator in the resolution of protein systems3,4.
Useful pH Range 6.1 - 7.5
Working concentration range 25mM-50mM7,8
Name CAS Molecular Formula Mol Wt Reagent Grades Life Science Grades PharmaGrade 
ACES 7365-82-4
C4H10N2O4S 182.2 A9758 RDD010 00194 A7949 A3594 RES1000A-A7
Interaction Comments
ACES binds Cu2+5. and Mg; weak interaction with Ca, Mn, Co, Ni and Zn6
Significant absorption of UV light at 230 nm5.
Literature References
  1. Good, N.E., et al, Hydrogen ion buffers for biological research. Biochemistry, 5(2), 467-477 (1966).
  2. Liu, Q., et al., pK-matched running buffers for gel electrophoresis. Anal. Biochem., 270(1), 112-122 (1999).
  3. Righetti, P.G., et al., Immobilized pH gradients: effect of salts, added carrier ampholytes and voltage gradients on protein patterns. Electrophoresis, 9(2), 65-73 (1988).
  4. Alonso, A., Human α-1-antitrypsin subtyping by hybrid isoelectric focusing in miniaturized polyacrylamide gel. Electrophoresis, 10(7), 513-519 (1989).
  5. Bollag, D.M. & Edelstein, S.J. (1992) Protein Methods. Chapter 1, II. Wiley-Liss. New York.
  6. Ferreira CM, Pinto IS, Soares, EV, Soares HM. 2015 Royal Society of Chemistry 30989- 31003.
  7. Zhou J., Methods in Molecular Medicine Microtubule protocols, Humana Press, 2010 pp 191.
  8. Stein R.L., Kinetics of Enzyme Action: Essential principles for drug hunters, Science 2011 pp 34.