Biological Buffers
Synonym: 4-(Cyclohexylamino)-1-butanesulfonic acid
Homolog of CHES and CAPS with higher pKa and similar utility in biological systems1.
Useful pH Range 10.0 - 11.4
Working concentration range No information found
        Comparison of products by grade or quality level
Name CAS Molecular Formula Mol Wt Reagent Grades Life Science Grades PharmaGrade  Solutions
CABS 161308-34-5 C10H21NO3S 235.34 C5580      
Interaction Comments
No reported interaction found
Literature References
  1. Thiel T, Liczkowski L, Bissen ST. J Biochem Biophys Methods. 1998 Nov 18;37(3):117-29.
  2. Ferreira CM, Pinto IS, Soares, EV, Soares HM. 2015 Royal Society of Chemistry 30989- 31003