Fast siRNA
Quick synthesis and shipment of custom siRNA using your sequences. U.S. orders must be received by 11 AM CST. European orders must be received by 11 AM CET.
Product Benefits
  • If you provide the sequences, your order will have a turnaround time of only 3 days
Product Features
  • If you have needs that are different from the general specifications presented here, please send a request to sirnarequest@sial.com.
Fast siRNA Specifications
Quantities
  • 2 (10 nmol) or 5 (25 nmol) OD
Purification
  • Desalt
Sequence Form
  • 19 to 23mer duplexes
Modifications
  • None
Quality Control
  • 100% mass spectrometry*
Format
  • Supplied in water in tubes
    • 50 µM (2 OD)
    • 100 µM (5 OD)
*Depending on manufacturing site, PAGE may be used to assess siRNA duplexes.
If additional help is needed, please consult our technical services group at oligotechserv@sial.com.