Custom miRNA
Custom miRNA synthesis using your sequences or sequences we have designed for you.
Product Benefits
  • Complimentary designs that mimic our Predesigned miRNA Mimics designs
  • Distinguish sequence-specific silencing from non–specific effects with 2 negative control miRNA
  • Custom services allow for flexibility in experimental set up
Product Features
  • For anything marked as ‘Inquire’ below or If you have needs that are different from the other general specifications presented, please send a request to sirnarequest@sial.com.
Custom miRNA Specifications
Quantities
  • 2 (10 nmol), 5 (25 nmol), 10 (50 nmol) & 50 (250 nmol) OD
  • For larger quantities & in-vivo-grade material, see our Custom iScale Oligos product
Purification
  • Desalt or HPLC
Sequence Form
  • 19 to 27mer duplexes
Modifications
  • Biotin, Phosphate, Amino-Modifier C7, 6-FAM™, Cyanine 3, Cyanine 5, Cyanine 5.5, 2’-OMe, Phosphorothioate & Thiol-Modifier-C6 S-S
Quality Control
  • 100% mass spectrometry**
Format
  • Supplied dry in tubes or plates
  • Pooling & aliquoting available (inquire for feasibility)
**Depending on manufacturing site, PAGE may be used to assess miRNA duplexes.
If additional help is needed, please consult our technical services group at oligotechserv@sial.com.