CRISPRi & CRISPRa Genetic Screening
Lentiviral Libraries and Pools for CRISPR Activation and Inhibition
Discover our optimized CRISPR activation and CRISPR inhibition libraries for gene knockdown and overexpression experiments. Our pooled CRISPRi and SAM CRISPRa lentiviral libraries are available off-the-shelf or customized. Individual guide RNAs for both systems are also easily ordered online.
Sigma-Aldrich® Advanced Genomics CRISPRi and CRISPRa libraries complement our comprehensive suite of genome engineering technologies to allow you to uncover further insights into complex biological processes. Large scale LOF (loss-of-function) and GOF (gain-of-function) screens can identify unique, yet functionally related, gene pathways and hits which are often missed under otherwise identical KO (knockout) screening parameters. Take your experiments Beyond the Bench with the largest genomics portfolio available and a partner that can ensure your experimental goals are met.
Sigma-Aldrich® CRISPRi and CRISPRa with dCas9 KRAB and dCas9 VP64
Figure 1: CRISPRi and CRISPRa utilize catalytically inactive dCas9 to recruit the transcriptional repressor KRAB domain or the transcriptional activators HSF1, p65 and MS2 to silence and overexpress target gene expression respectively.
Specifications
          CRISPRi           CRISPRa
  • ~10 gRNAs/gene
  • ~258,000 total clones
  • Targets the human genome
  • Lentivirus provided as 200 µL (8 x 25 µuL) aliquots at a minimum of 5x108 TU/mL
  • Off-the-shelf subpools available
  • ~3 gRNAs/gene
  • ~70,000 total clones
  • Targets human and mouse genomes
  • Lentivirus provided as 200 µL (8 x 25 uµLl) aliquots at a minimum of 5x108 VP/mL
  • Custom subpools available
CRISPRi Available Libraries and Pools
Product Description Target Number of Genes Product Number
Whole-Genome Libraries
Human Whole Genome & Non-Coding CRISPRi Library
Human, Whole genome
18,905
CRISPRILIB
Control & Effector Kit: Positive & Negative controls with dCas9
Coding & non-targeting
Coding gene RAB1A and a non-targeting control
CRISPRICON
CRISPRi Subpools
10x Genomics Feature Barcode Optimization Kit 
Human, Coding Coding gene RAB1A and a non-targeting control
CRISPRI10X
Druggable Genome Pool 
Human, Coding 2,318
CRISPRIS01
Cancer & Apoptosis Pool   Human, Coding 2,916
CRISPRIS02
Stress Proteostasis Pool
Human, Coding> 3,093 CRISPRIS03
Mitochondria/Trafficking/Motility Pool   Human, Coding 2,219
CRISPRIS04
Gene Expression Pool (Transcription Factors)
Human, Coding 2,292 CRISPRIS05
Membrane Proteins Pool   Human, Coding 2,418 CRISPRIS06
Unassigned Pool (Uncategorized Gene Function, Novel Targets)
Human, Coding 1,824 CRISPRIS07
Common Cancer Cell Lines Pool & iPSC Pool: Common to 7 cell lines (5 cancer, iPSC, HFF)
Human, Long
non-coding
691 CRISPRIS08
iPSC Pool 
Human, Long
non-coding
3,000
CRISPRIS09
Common Cancer Cell Lines Pool (HeLa, HEK293T, K562, MCF7, U87 cell lines) Human, Long
non-coding
1,329
CRISPRIS10
SAM CRISPRa Libraries and Helper Constructs
Product Description Target Number of Genes Product Number
Whole-Genome SAM Libraries
SAM CRISPRa Library Zeo 
Human, coding
23,430
HSAMZEO
SAM CRISPRa Library Puro
Human, coding
23,430 HSAMPURO
SAM CRISPRa Library Zeo
Mouse, coding
23,439 MSAMZEO
SAM CRISPRa Library Puro
Mouse, coding
23,439 MSAMPURO
SAM Helper Constructs: Plasmid DNA
dCas9-VP64-Blasticidin SAM Helper Construct 1
Mammalian   SAMVP64BST
MS2-P65-HSF1-Hygromycin SAM Helper Construct 2
Mammalian   SAMMS2HYG
SAM Helper Construct Kit
Mammalian   SAMHELPERP
SAM Helper Constructs: Lentivirus
dCas9-VP64-Blasticidin SAM Helper Construct 1 Mammalian   SAMVP64BSTV
MS2-P65-HSF1-Hygromycin SAM Helper Construct 2
Mammalian   SAMMS2HYGV
SAM Helper Construct Kit
Mammalian   SAMHELPERV
Not sure which screening tool is right for your experiment? Pricing questions? Need a custom pool? We can help!
   CONTACT US
Resources
The Sigma-Aldrich® CRISPR inhibition libraries have superior algorithm placement and improved sgRNA scaffolding for optimal gene knockdown, even in the most difficult targets. 
Added Benefits:
Vector Maps
SAM CRISPRa
CRISPRi
Related Links