The MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A is a multiplex assay that combines tests for 48 individual immune factors that previously have not been together in a single panel, while also allowing you to choose any combination of analytes, or select a 48-plex or a 38-plex premixed kit. This panel was built with quality in mind and has improved workflow and sample detection that help you discover with confidence.
Figure 1. 48-plex standard curves were performed in the MXHSM-A serum matrix except *RANTES, which was performed in the L-AB assay buffer. The kit was run using the MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A (Cat. No. HCYTA-60K) overnight assay protocol.
What is a Multiplex Assay?
A multiplex assay is an assay that combines tests for various analytes in a single panel. These assays are gaining popularity for their advantages of saving sample volume and time by improving workflow when compared to assays that measure a single analyte, such as enzyme-linked immunosorbent assays (ELISA). Our MILLIPLEX® multiplex assays are the most highly verified multiplex assays for Luminex® technology. They can measure over 1,000 analytes including cytokines, chemokines, and growth factors. Discover more about the benefits of MILLIPLEX® multiplex immunoassays in this short video.
Role of Cytokines, Chemokines, and Growth Factors
Cytokines mediate direct interactions between cells and regulate processes taking place in the extracellular environment. Cytokines differ from hormones in that they act on a wider spectrum of target cells and include lymphokines, interferons, colony-stimulating factors, and chemokines. Additionally, growth factors are involved in the stimulation of target cell survival, proliferation, and differentiation with effects on angiogenesis, vasculogenesis, and cell migration.
Cytokine, chemokine, and growth factor research plays a significant role in achieving a deeper understanding of the immune system and its multi-faceted response to most antigens. This deep understanding is especially relevant to those responses that make up the inflammatory process and disease states, such as osteoarthritis, IBD, sepsis, allergic reactions, and even cancer. It also allows researchers to understand infectious, respiratory, neurologic, metabolic, and cardiovascular diseases. Explore how MILLIPLEX® multiplex assays can help to understand cytokine storm in our MILLIPLEX® Multiplex Assays for Research Applications in COVID-19 (SARS-CoV-2) article. For Research Use Only. Not For Use In Diagnostic Procedures.
Quality Built into MILLIPLEX® Kits
Kit development and verification entails testing for selectivity and specificity to ensure negligible cross-reactivity in the tested sample types, as well as assay specificity to ensure consistent performance of an assay in single-plex vs. multiplex formats. Buffers and diluents are optimized to enhance antibody specificity, such that only those analytes of interest are detected in samples. The serum matrix is also carefully selected and optimized for use in the standard curve when using serum or plasma samples to most closely mimic sample matrix, thus normalizing assay performance. The streptavidin-phycoerythrin (SAPE) concentration is titrated in-house for optimal signal and is provided ready-to-use with no dilution required. Additionally, all our kits are rigorously tested for shipping stability, and samples are also tested for temperature and freeze/thaw tolerance.
Sample dilution is optimized for each analyte in the kit, ensuring that biologically relevant sample values are detectable, and fall within the dynamic range of the standard curves. Samples tested for this kit include normal and disease serum and plasma samples, as well as peripheral blood mononuclear cell supernatants (PBMCs, unstimulated and stimulated with various agents). Quality Controls (QCs), which are low and high dilutions of recombinant proteins for each analyte, are manufactured such that the two QCs (high and low) have optimal placement on each standard curve. QC range sheets are provided with each kit. Additionally, we always recommend users include experiment-specific samples for use as controls in each assay. This quick video describes why our high-quality MILLIPLEX® multiplex assays guarantee reliable performance across lots.
Workflow Improvements
An assay containing 48 individual immune factors in a single panel is a powerful tool for research. The ability to select all analytes, perhaps for screening, and then to select a subset of analytes as a project continues, allows researchers the flexibility they need to work efficiently. Detecting all (or a subset) of these proteins at pg/mL levels within a kit with standard curves that do not change from lot to lot makes the assay very user-friendly. Consistent standard curves from lot to lot ensure consistent sample results across a project. The MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A also has a straightforward protocol with a familiar workflow in which every step is outlined to ensure ease of use. Learn about the MILLIPLEX® protocol in our How to Run MILLIPLEX® Multiplex Immunoassays video.
Another assay improvement is that both types of platelet-derived growth factor PDGF-AA and PDGF-AB/BB can now be tested with neat serum and plasma samples, so there is no need for sample dilution as is the case with MILLIPLEX® Human Cytokine/Chemokine Panel 1 (HCYTOMAG-60K), or for other Luminex® based kits on the market. RANTES, being highly expressed in serum and plasma samples, still requires a separate sample dilution in serum and plasma of 1:100.
A simple assay improvement of logically arranging the bead regions and analyte names in numeric-alphabetic order, allows researchers to quickly locate specific analytes in the result output files much easier than with previous assays. This helps immensely when analyzing datasets of 42 analytes or greater.
Sample Detectability Using MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A (Cat. No. HCYTA-60K)
In addition to improvements in the standard curves relative to the comparative analyte assays for the MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A, there is also improvement in analyte performance in terms of sample detection. One goal in the development of this panel was to closely match sample values when evaluated against comparison MILLIPLEX® kits (Cat. No. HCYTOMAG-60K, HCYP3MAG-63K, or HTH17MAG-14K). Of note, three of the analyte assays have been adjusted to better match the accepted values in the literature: GROα1,2, IL-41, and IL-223,4,5.
Figure 2. Testing disease samples and stimulated PBMC using the MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A (Cat. No. HCYTA-60K).
More details can be found in our MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A application note.
MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A Products
Cat. No. |
Number of Analytes |
Analytes Contained |
Notes |
---|
HCYTA-60K1 |
Up to 48 |
sCD40L, EGF, Eotaxin, FGF-2, FLT-3L, Fractalkine, G-CSF, GM-CSF, GROα, IFNα2, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12 (p40), IL-12 (p70), IL-13, IL-15, IL-17A, IL-17E/IL-25, IL-17F, IL-18, IL-22, IL-27, IP-10, MCP-1, MCP-3, M-CSF, MDC, MIG, MIP-1α, MIP-1β, PDGF-AA, PDGF-AB/BB, RANTES*, TGFα, TNFα, TNFβ, VEGF-A |
1Customizable panel: choose your analytes and receive individual bead vials or use our convenient service for customizing your own bead premix (additional charge applicable). |
HCYTA-60K-PX382,
HCYTA-60K-PXBK383 |
38 |
sCD40L, EGF, Eotaxin, FGF-2, FLT-3L, Fractalkine, G-CSF, GM-CSF, GROα, IFNα2, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12 (p40), IL-12 (p70), IL-13, IL-15, IL-17A, IL-17E/IL-25, IL-17F, IL-18, IL-22, IL-27, IP-10, MCP-1, MCP-3, M-CSF, MDC, MIG, MIP-1α, MIP-1β, PDGF-AA, PDGFAB/BB, RANTES*, TGFα, TNFα, TNFβ, VEGF-A |
2Premix kits contain either a 37- or 47-plex bead set, plus a vial of RANTES beads.
3Bulk kits match the formatting of Premix kits but arrive in Space Saver packaging. |
HCYTA-60K-PX482, HCYTA-60K-PXBK483 |
48 |
sCD40L, EGF, Eotaxin, FGF-2, FLT-3L, Fractalkine, G-CSF, GM-CSF, GROα, IFNα2, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12 (p40), IL-12 (p70), IL-13, IL-15, IL-17A, IL-17E/IL-25, IL-17F, IL-18, IL-22, IL-27, IP-10, MCP-1, MCP-3, M-CSF, MDC, MIG, MIP-1α, MIP-1β, PDGF-AA, PDGF-AB/BB, RANTES*, TGFα, TNFα, TNFβ, VEGF-A |
*RANTES is always provided as a separate bead vial due to different dilution requirements for serum/ plasma samples. If measuring RANTES in serum/plasma, it is recommended to use a single-plex kit including RANTES only. |
For Research Use Only. Not For Use In Diagnostic Procedures.
MILLIPLEX® kits are manufactured in facilities which are ISO 9001:2015 compliant and are for research use only (RUO), not for use in clinical or for medical diagnostic purposes.
View our large portfolio of MILLIPLEX® assays for Luminex® technology at SigmaAldrich.com/milliplex.
Find answers to common questions about MILLIPLEX® assays in our MILLIPLEX® Multiplex Assays for Luminex® FAQs page.
Recent Publications Using MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A (Cat. No. HCYTA-60K)
- Shi, Hongbo, Wenjing Wang, Jiming Yin, Yabo Ouyang, Lijun Pang, Yingmei Feng, Luxin Qiao, et al. 2020. “The Inhibition of IL-2/IL-2R Gives Rise to CD8+T Cell and Lymphocyte Decrease through JAK1-STAT5 in Critical Patients with COVID-19 Pneumonia.” Cell Death & Disease 11(6). https://doi.org/10.1038/s41419-020-2636-4.
- Leng, Zikuan, Rongjia Zhu, Wei Hou, Yingmei Feng, Yalei Yang, Qin Han, Guangliang Shan, et al. 2020. “Transplantation of ACE2 - Mesenchymal Stem Cells Improves the Outcome of Patients With COVID-19 Pneumonia.” Aging and Disease 11 (2): 216–228.https://doi.org/10.14336/ad.2020.0228.
- Ouyang, Yabo, Jiming Yin, Wenjing Wang, Hongbo Shi, Ying Shi, Bin Xu, Luxin Qiao, et al. 2020. “Down-Regulated Gene Expression Spectrum and Immune Responses Changed during the Disease Progression in COVID-19 Patients.” Clinical Infectious Diseases. https://doi.org/10.1093/cid/ciaa462.
- Fraser, Douglas D., Gediminas Cepinskas, Marat Slessarey, Claudio Martin, Mark Daley, Michael R. Miller, David B. O’Gordman, et al. 2020. “Inflammation Profiling of Critically Ill Coronavirus Disease 2019 Patients.” Critical Care Explorations 2 (6): e0144. https://doi.org/10.1097/CCE.0000000000000144.
References
- Kleiner, Giulio, Annalisa Marcuzzi, Valentina Zanin, Lorenzo Monasta, and Giorgio Zauli. 2013. “Cytokine Levels in the Serum of Healthy Subjects.” Mediators of Inflammation 2013: 1–6. https://doi.org/10.1155/2013/434010.
- Mitsuyama, Keiichi, Osamu Tsuruta, Nobuo Tomiyasu, Kosuke Takaki, Asuka Suzuki, Junya Masuda, Hiroshi Yamasaki, Atsushi Toyonaga, and Michio Sata. 2006. “Increased Circulating Concentrations of Growth-Related Oncogene (GRO)-α in Patients with Inflammatory Bowel Disease.” Digestive Diseases and Sciences 51 (1): 173–77. https://doi.org/10.1007/s10620-006-3104-4.
- Lin, J., L. H. Yue, and W. Q. Chen. 2014. “Decreased Plasma IL-22 Levels and Correlations with IL-22-Producing T Helper Cells in Patients with New-Onset Systemic Lupus Erythematosus.” Scandinavian Journal of Immunology 79 (2): 131–36. https://doi.org/10.1111/sji.12135.
- Herder, Christian, Julia M. Kannenberg, Maren Carstensen-Kirberg, Cornelia Huth, Christa Meisinger, Wolfgang Koenig, Annette Peters, Wolfgang Rathmann, Michael Roden, and Barbara Thorand. 2017. “Serum Levels of Interleukin-22, Cardiometabolic Risk Factors and Incident Type 2 Diabetes: KORA F4/FF4 Study.” Cardiovascular Diabetology 16 (1). https://doi.org/10.1186/s12933-017-0498-6.
- Oliveira, Priscilla Stela Santana de, Pablo Ramon Gualberto Cardoso, Emerson Vasconcelos de Andrade Lima, Michelly Cristiny Pereira, Angela Luzia Branco Pinto Duarte, Ivan da Rocha Pitta, Moacyr Jesus Barreto de Melo Rêgo, and Maira Galdino da Rocha Pitta. 2015. “IL-17A, IL-22, IL-6, and IL-21 Serum Levels in Plaque-Type Psoriasis in Brazilian Patients.” Mediators of Inflammation 2015: 1–5. https://doi.org/10.1155/2015/819149.